Skip to Content
Merck
All Photos(1)

Key Documents

95446

Supelco

1-Octanol

analytical standard

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

4.5 (vs air)

vapor pressure

0.14 mmHg ( 25 °C)

Assay

≥99.7% (GC)

autoignition temp.

523 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.429 (lit.)
n20/D 1.429

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Octanol is saturated fatty alcohol with potential application as a biofuel due to its diesel-like properties.[1]
1-Octanol may be used in combination with methanol as the mobile phase, to obtain a lipophilicity index for a set of complex molecules, using reversed phase high performance liquid chromatography method (RP-HPLC).[2]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

1-Octanol may be used as an extraction solvent in the determination of ochratoxin A[3] and neonicotinoid insecticide residues,[4] wine samples,[3] honey samples,[4] and tea samples[5] using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dynamic hollow fiber protected liquid phase microextraction and quantification using gas chromatography combined with electron capture detection of organochlorine pesticides in green tea leaves and ready-to-drink tea
Huang P-S and Huang D-S
Journal of Chromatography A, 1135(1), 6-11 (2006)
In-coupled syringe assisted octanol?water partition microextraction coupled with high-performance liquid chromatography for simultaneous determination of neonicotinoid insecticide residues in honey
Vichapong J, et al.
Talanta, 139(2), 21-26 (2015)
Determination of lipophilicity by reversed-phase high-performance liquid chromatography: Influence of 1-octanol in the mobile phase
Liu X, et al.
Journal of Chromatography A, 1091(1), 51-59 (2005)
Determination of ochratoxin A in wine using liquid-phase microextraction combined with liquid chromatography with fluorescence detection
Gonzalez-Penas E, et al.
Journal of Chromatography A, 1025(2), 163-168 (2004)
Microbial production of 1-octanol: a naturally excreted biofuel with diesel-like properties.
Akhtar MK, et al.
Metabolic Engineering Communications, 2, 1-5 (2015)

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service