Merck
  • Modification of a gas chromatography/atmospheric pressure chemical ionisation time-of-flight mass spectrometer as an alternative to automated atmospheric pressure solids analysis probe.
Modification of a gas chromatography/atmospheric pressure chemical ionisation time-of-flight mass spectrometer as an alternative to automated atmospheric pressure solids analysis probe.
Rapid communications in mass spectrometry : RCM (2014-08-19)
Peter Stokes, David Parker, Jackie Mosely
ABSTRACT
The atmospheric pressure solids analysis probe (ASAP) has been successfully introduced into laboratories as a routine analytical tool but the technique is labour-intensive and is not easily automated. This paper describes the modifications made to an existing atmospheric pressure gas chromatography (APGC) system to enable the automated analysis of samples that would have otherwise been analysed by ASAP. Liquid samples were injected into a 1 m piece of fused silica guard column using a standard split/splitless gas chromatography (GC) inlet, oven and autosampler. A novel timing delay circuit was developed and integrated into the system to control acquisition start times and a bespoke heating block/calibrant delivery system was used to transport either the calibration or reference compound into the ionisation source. The modified system has allowed the successful analysis of those samples requiring ASAP to be carried out in a fully automated manner using APGc. The analysis is rapid and can be carried out in less than 2 min. The results obtained are directly comparable with those obtained by ASAP. A typical mass accuracy of better than 5 mm/z units was achieved, allowing the molecules to be identified based on their elemental formulae. An existing APGC system has been successfully modified and is suitable for the automatic analysis of samples normally analysed by ASAP. The APGC instrument incorporates the use of a novel reference compound and delivery system which allows accurate mass measurements to be performed. This and the other modifications described have allowed the technique to be incorporated into an existing suite of mass spectrometry-based experiments for use in the characterisation of organic molecules.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Tetraiodoethylene, 97%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Supelco
Melting point standard 47-49°C, analytical standard
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Leucine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%