Skip to Content
Merck
All Photos(3)

Key Documents

W332615

Sigma-Aldrich

Acetone

greener alternative

natural, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
FEMA Number:
3326
Beilstein:
635680
EC Number:
Council of Europe no.:
737
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.050
NACRES:
NA.21

grade

natural

reg. compliance

FDA 21 CFR 117

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥97%

form

liquid

expl. lim.

13.2 %

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

clear colorless

refractive index

n20/D 1.359 (lit.)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

format

neat

greener alternative category

Organoleptic

apple; ethereal

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone is a volatile liquid mainly used as a solvent, color diluent and flavor enhancer in the food industry. It is also used as an extractant due to its miscibility with many organic solvents, water, and oils.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is BioRenewable and thus aligns with "Designing Safer Chemicals" and "Use of Renewable Feedstocks".

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Handbook of Preservatives, 267-267 (2004)
Encyclopedia of Food and Color Additives, 1, 20-22 (1997)
Oleksiy Krupin et al.
Optics express, 21(1), 698-709 (2013-02-08)
Straight long-range surface plasmon waveguides are demonstrated as biosensors for the detection of cells, proteins and changes in the bulk refractive index of solutions. The sensors consist of 5 μm wide 22 nm thick Au stripes embedded in polymer (CYTOP™)
Len Verbeke et al.
Hepatology (Baltimore, Md.), 59(6), 2286-2298 (2013-11-22)
The farnesoid X receptor (FXR) is a nuclear bile acid receptor involved in bile acid homeostasis, hepatic and intestinal inflammation, liver fibrosis, and cardiovascular disease. We studied the effect of short-term treatment with obeticholic acid (INT-747), a potent selective FXR
Wanqiang Sheng et al.
Cell research, 24(12), 1387-1402 (2014-11-22)
T helper (TH)-cell subsets, such as TH1 and TH17, mediate inflammation in both peripheral tissues and central nervous system. Here we show that STAT5 is required for T helper-cell pathogenicity in autoimmune neuroinflammation but not in experimental colitis. Although STAT5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service