Skip to Content
Merck
All Photos(1)

Key Documents

01-0530

Sigma-Aldrich

Acetone

for chromatography, ≥99.8%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

for chromatography

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.8%

form

liquid

expl. lim.

13.2 %

availability

available only in Japan

refractive index

n20/D 1.359 (lit.)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleksiy Krupin et al.
Optics express, 21(1), 698-709 (2013-02-08)
Straight long-range surface plasmon waveguides are demonstrated as biosensors for the detection of cells, proteins and changes in the bulk refractive index of solutions. The sensors consist of 5 μm wide 22 nm thick Au stripes embedded in polymer (CYTOP™)
Len Verbeke et al.
Hepatology (Baltimore, Md.), 59(6), 2286-2298 (2013-11-22)
The farnesoid X receptor (FXR) is a nuclear bile acid receptor involved in bile acid homeostasis, hepatic and intestinal inflammation, liver fibrosis, and cardiovascular disease. We studied the effect of short-term treatment with obeticholic acid (INT-747), a potent selective FXR
Wanqiang Sheng et al.
Cell research, 24(12), 1387-1402 (2014-11-22)
T helper (TH)-cell subsets, such as TH1 and TH17, mediate inflammation in both peripheral tissues and central nervous system. Here we show that STAT5 is required for T helper-cell pathogenicity in autoimmune neuroinflammation but not in experimental colitis. Although STAT5
Hugo Lövheim et al.
Alzheimer's & dementia : the journal of the Alzheimer's Association, 11(6), 593-599 (2014-07-22)
Previous studies have suggested a link between herpes simplex virus (HSV) type 1 and the development of Alzheimer's disease (AD). The present analysis included 3432 persons (53.9% women, mean age at inclusion 62.7 ± 14.4 years) with a mean follow-up
Juan A López-Ráez et al.
Plant science : an international journal of experimental plant biology, 230, 59-69 (2014-12-07)
Apocarotenoids are a class of compounds that play important roles in nature. In recent years, a prominent role for these compounds in arbuscular mycorrhizal (AM) symbiosis has been shown. They are derived from carotenoids by the action of the carotenoid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service