Skip to Content
Merck
All Photos(3)

Key Documents

179124

Sigma-Aldrich

Acetone

ACS reagent, ≥99.5%

Synonym(s):

2-propanone, Dimethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12352115
eCl@ss:
39021201
PubChem Substance ID:
NACRES:
NA.02

grade

ACS reagent

Quality Level

Agency

suitable for EPA 1613

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

shelf life

Recommended retest period - 2 years

expl. lim.

13.2 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0003 meq/g Titr. acid
≤0.0006 meq/g Titr. base
≤0.002% aldehyde as formaldehyde
≤0.05% isopropanol
≤0.05% methanol
≤0.5% water

evapn. residue

≤0.001%

color

APHA: ≤10
clear

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

format

neat

storage temp.

room temp

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone is a polar, aprotic organic solvent. It is a colorless and highly volatile liquid. It is commonly used in organic syntheses as solvent and/or building block, in pharmaceutical and personal care industries. This grade meets ACS requirements, making it ideal for a wide range of analytical applications.

Application

  • Used as solvent and/or building block in organic syntheses, such as SN2 reactions and Jones oxidation
  • Mobile phase in Thin layer chromatography (TLC)
  • Used as solvent in chemical molecules delivery systems in pharmaceutical and cosmetic industries
  • Cleaning proposes
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Features and Benefits

ACS solvents meet or exceed the high standards of the American Chemical Society (ACS) ,with test specifications that are specialized to every compound. According to the American Chemical Society, ACS reagent grade implies that it is a substance of sufficient purity to be used in most chemical analyses or reactions.

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Legal Information

Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of 1,2-diketones: oxidation of alkynes by potassium permanganate in aqueous acetone
Srinivasan NS and Donald GL
The Journal of Organic Chemistry, 44(9), 1574-1574 (1979)
Aldol condensation of acetone over layered double hydroxides of the meixnerite type.
Tichit D, et al.
Applied Clay Science, 13(5), 401-415 (1998)
An investigation on photocatalytic activities of mixed TiO2-rare earth oxides for the oxidation of acetone in air.
Lin J and Jimmy CY.
Journal of Photochemistry and Photobiology A: Chemistry, 116(1), 63-67 (1998)
Oleksiy Krupin et al.
Optics express, 21(1), 698-709 (2013-02-08)
Straight long-range surface plasmon waveguides are demonstrated as biosensors for the detection of cells, proteins and changes in the bulk refractive index of solutions. The sensors consist of 5 μm wide 22 nm thick Au stripes embedded in polymer (CYTOP™)
Wanqiang Sheng et al.
Cell research, 24(12), 1387-1402 (2014-11-22)
T helper (TH)-cell subsets, such as TH1 and TH17, mediate inflammation in both peripheral tissues and central nervous system. Here we show that STAT5 is required for T helper-cell pathogenicity in autoimmune neuroinflammation but not in experimental colitis. Although STAT5

Articles

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Protocols

Derived from procedure SSPHYT02. Includes template updates to current SOP specifications and incorporation of notes into the procedure.

Derived from procedure SSPHYT02. Includes template updates to current SOP specifications and incorporation of notes into the procedure.

Derived from procedure SSPHYT02. Includes template updates to current SOP specifications and incorporation of notes into the procedure.

Derived from procedure SSPHYT02. Includes template updates to current SOP specifications and incorporation of notes into the procedure.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service