Merck
  • Embedment of Ag(I)-organic frameworks into silica gels for microextraction of polybrominated diphenyl ethers in soils.
Embedment of Ag(I)-organic frameworks into silica gels for microextraction of polybrominated diphenyl ethers in soils.
Journal of chromatography. A (2015-01-27)
Chao-Yan Zhang, Zeng-Guang Yan, You-Ya Zhou, Lin Wang, Ya-Bo Xie, Li-Ping Bai, Hui-Yan Zhou, Fa-Sheng Li
ABSTRACT
Three Ag(I)-organic frameworks, [Ag5(pydc)2(CN)]n, {[Ag4(pydc)2]CH3CN}n, and [Ag(4,4'-bpy)NO3]n, were synthesized and embedded into silica gels to form metal-organic-framework (MOF)-embedded gels for the microextraction of polybrominated diphenyl ethers (PBDEs) in soils. Despite the great differences in the structures of the organic ligands, all three Ag(I)-organic frameworks were found to effectively accumulate and concentrate PBDEs from sample solutions prepared with contaminated soil and purified water, indicating the important roles of Ag centers in PBDE extraction. Under the optimal experimental conditions (MOF mass, water volume, temperature, extraction time, and back-extraction time) for PBDE extraction from sample solutions, the detection limits of seven PBDEs (BDE-28, 47, 99, 100, 153, 154, and 183) ranged from 0.01 to 2.6ngg(-1) for [Ag5(pydc)2(CN)]n, 0.20-0.64ngg(-1) for {[Ag4(pydc)2]CH3CN}n, and 0.60-3.08ngg(-1) for [Ag(4,4'-bpy)NO3]n. The reproducibilities of the three methods were all satisfactory with relative standard deviations (RSDs) in the range of 2.2-9.6%, 5.3-10.4%, and 6.9-9.4% for [Ag5(pydc)2(CN)]n, {[Ag4(pydc)2]CH3CN}n, and [Ag(4,4'-bpy)NO3]n, respectively. The use of Ag(I)-organic frameworks for the microextraction of PBDEs was validated using both certified reference soils and field-contaminated soils, and the proposed methods are recommended as rapid and environmentally friendly alternatives for the extraction and determination of PBDEs in soils.
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoroacetic acid, SAJ special grade, ≥99.0%
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
Toluene, analytical standard
Supelco
Toluene, HPLC grade, 99.8%
Supelco
2,2,4-Trimethylpentane, analytical standard
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
2,2,4-Trimethylpentane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%