Skip to Content
Merck
All Photos(1)

Key Documents

PHR1317

Supelco

Toluene

Residual Solvent, Pharmaceutical Secondary Standard, Certified Reference Material

Synonym(s):

Toluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1601805

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

CofA

current certificate can be downloaded

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-30°C

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Residual Solvent – Toluene is a certified reference material for the screening and quantification of residual solvents. This CRM is a secondary pharmaceutical reference standard and is traceable and qualified against the corresponding pharmacopeial standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1709 in the slot below. This is an example certificate only and may not be the lot that you receive.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elke Persch et al.
ChemMedChem, 9(8), 1880-1891 (2014-05-03)
The causative agents of the parasitic disease human African trypanosomiasis belong to the family of trypanosomatids. These parasitic protozoa exhibit a unique thiol redox metabolism that is based on the flavoenzyme trypanothione reductase (TR). TR was identified as a potential
Murat Yücel et al.
Neuroscience and biobehavioral reviews, 32(5), 910-926 (2008-05-06)
Organic solvent abuse is associated with increased risk for serious medical, neurological, and neuropsychological impairments. While animal research suggests that exposure to organic solvents (especially toluene) may be neurotoxic, much less is known about the consequences of long-term exposure in
Yoko Yamakoshi et al.
Faraday discussions, 173, 287-296 (2014-12-04)
The photoinduced reactive oxygen species (ROS) generation from several water-soluble fullerenes was examined. Macromolecular or small molecular water-soluble fullerene complexes/derivatives were prepared and their (1)O(2) and O(2)˙(-) generation abilities were evaluated by ESR spin-trapping methods. As a result, efficient (1)O(2)
Perrine Hoet et al.
Critical reviews in toxicology, 38(2), 127-170 (2008-02-09)
Noise is the most common preventable cause of irreversible sensorineural hearing loss. During recent years, the results of experimental and human investigations have raised the level of concern about the potential ototoxicity of chemical agents and their interaction with noise.
Emma Oakton et al.
Dalton transactions (Cambridge, England : 2003), 43(40), 15138-15142 (2014-09-03)
Herein, we report the preparation of small and narrowly distributed (2.1 ± 0.5 nm) Ag nanoparticles supported on passivated silica, where the surface OH groups are replaced by OSiMe3 functionalities. This synthetic method involves the grafting of silver(I) bis(trimethylsilyl)amide ([AgN(SiMe3)2]4)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service