Skip to Content
Merck
All Photos(2)

Documents

57362-U

Supelco

SPME Fiber Assortment Kit

greener alternative

DVB/CAR/PDMS, PDMS/DVB, PDMS, CAR/PDMS, for use with autosampler, Nitinol-core (NIT), 23 ga, 65 μm (PDMS/DVB), 100 μm (PDMS), 50 μm (DVB layer in DVB/CAR/PDMS), 30 μm (CAR/PDMS layer in DVB/CAR/PDMS), 85 μm (CAR/PDMS)

Synonym(s):

Solid Phase Microextraction Fiber Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

product name

SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler

material

assorted colors hub
fused silica fiber

Quality Level

needle size

23 ga

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

100 μm (PDMS)
30 μm (CAR/PDMS layer in DVB/CAR/PDMS)
50 μm (DVB layer in DVB/CAR/PDMS)
65 μm (PDMS/DVB)
85 μm (CAR/PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

CAR/PDMS coating
DVB/CAR/PDMS coating
PDMS coating
PDMS/DVB coating

application(s)

food and beverages

compatibility

for use with autosampler

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Components

SPME Fiber Assortment Kit 5 contains one fiber of each:
  • 65μm PDMS/DVB coating
  • 50/30μm DVB/Carboxen/PDMS coating
  • 85μm Carboxen/PDMS coating
  • 100μm PDMS coating

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Tytgat et al.
International journal of legal medicine, 109(3), 150-154 (1996-01-01)
The most common alkyl nitrites encountered in forensic toxicology are iso-butyl, n-butyl and iso-pentyl(amyl) nitrites. All have become popular as an aphrodisiac, especially among the homosexual population. Alkyl nitrites are a volatile and unstable group of compounds, which hydrolyse in
Jermiah Y Shen et al.
Journal of separation science, 35(19), 2544-2552 (2012-08-22)
The presence of triclosan and triclocarban, two endocrine-disrupting chemicals and antimicrobial agents, and transformation products of triclocarban, 1,3-di(phenyl)urea, 1,3-bis(4-chlorophenyl)urea and 1,3-bis(3,4-dichlorophenyl)urea, in tap water, treated household drinking water, bottled water, and river water samples were investigated using solid-phase micro-extraction coupled
A P Arisseto et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 29(12), 1832-1839 (2012-08-23)
Selected commercial processed foods available in the Brazilian market (306 samples) were analysed for furan content using a validated gas chromatography-mass spectrometry method preceded by headspace solid phase micro-extraction (HS-SPME-GC/MS). Canned and jarred foods, including vegetable, meat, fruit and sweet
Alfredo Koch et al.
Phytochemistry, 71(17-18), 2190-2198 (2010-10-23)
2-Methoxy-3-isobutylpyrazine (MIBP) contributes a bell pepper aroma to many grape cultivars and has a reported aroma threshold of ∼2 ng L(-1) in water. The purpose of this study was twofold: (1) develop a procedure using headspace solid phase micro-extraction combined
Cristina Sala et al.
Journal of agricultural and food chemistry, 52(11), 3492-3497 (2004-05-27)
The influence of vine training and sunlight exposure on the 3-alkyl-2-methoxypyrazines contents in musts and wines was studied by means of two previously reported methods based on headspace solid-phase micro-extraction. Experimental samples were monitored throughout grape ripening and wine making.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service