Skip to Content
Merck
All Photos(1)

Key Documents

89680

Supelco

Toluene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12191501
eCl@ss:
39011102
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

≥99.9% (GC)

autoignition temp.

997 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene is a volatile organic compound and a priority environmental contaminant as designated by the US EPA.[1] The effect of toluene on the central nervous system myelin has been studied.[2]
Toluene is an aromatic hydrocarbon[3], which can be obtained from crude oil via cracking or distillation process.[4]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Toluene may be used as an analytical reference standard for the quantification of the analyte in water samples[1] and engine oils[5] using gas chromatography (GC) with flame ionization detection (FID)[1] and mass spectrometry (MS).[5]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformation of toluene and benzene by mixed methanogenic cultures
Grbic-galic and Vogel.MT
Applied and Environmental Microbiology, 53(2), 254-260 (1987)
Tang W and Eisenbrand G et al.
Chinese Drugs of Plant Origin: Chemistry, Pharmacology, and Use in Traditional and Modern Medicine (2013)
Headspace microdrop analysis?an alternative test method for gasoline diluent and benzene, toluene, ethylbenzene and xylenes in used engine oils
Kokosa MJ and Przyjazny A
Journal of Chromatography A, 983(1-2), 205-214 (2003)
Determination of benzene, toluene, ethylbenzene and xylene in river water by solid-phase extraction and gas chromatography
Mottaleb AM, et al.
Analytical Sciences, 19(10), 1365-1369 (2003)
Nordic Council of Ministers et al.
Effects on Reproduction of Styrene, Toluene and Xylene (1990)

Articles

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service