Skip to Content
Merck
All Photos(1)

Key Documents

F13002

Sigma-Aldrich

3-Fluorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4OH
CAS Number:
Molecular Weight:
112.10
Beilstein:
1904536
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.514 (lit.)

bp

178 °C (lit.)

mp

8-12 °C (lit.)

density

1.238 g/mL at 25 °C (lit.)

SMILES string

Oc1cccc(F)c1

InChI

1S/C6H5FO/c7-5-2-1-3-6(8)4-5/h1-4,8H

InChI key

SJTBRFHBXDZMPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao-jie Zhang et al.
Huan jing ke xue= Huanjing kexue, 27(9), 1841-1845 (2006-11-30)
An acclimated activated sludge was examined for its ability to degrade 3-fluorophenol in aerobic batch cultures. The result indicated that the organism degrades up to 100 mg/L 3-fluorophenol completely with approximately 100% fluoride anion release within 16 h. 3-Fluorophenol can
Zhang Chaojie et al.
Journal of hazardous materials, 141(1), 295-300 (2006-08-16)
An acclimated activated sludge was examined for its ability to degrade meta-fluorophenol as sole carbon source in aerobic batch cultures. The mechanism study revealed that the initial step in the aerobic biodegradation of meta-fluorophenol was their transformation to fluorocatechol. Following
S Peelen et al.
European journal of biochemistry, 218(2), 345-353 (1993-12-01)
The regioselectivity and rate of the ortho-hydroxylation of 3-fluorophenol by phenol hydroxylase from Trichosporon cutaneum (EC 1.14.13.7) was studied using 19F-NMR. The regioselective hydroxylation as well as the rate of ortho-hydroxylation are pH dependent with a pKa of 6.5. At
Dawn Marie Reinhold et al.
Environmental toxicology and chemistry, 25(11), 3039-3045 (2006-11-09)
The effects of contaminant concentration on contaminant removal by Lemna minor were examined under elevated plant densities typical of full surface coverage. Uptake of 3-fluorophenol and 3-trifluoromethylphenol by L. minor was determined at concentrations from 10 to 1750 microM. Because
Norihiro Tsuji et al.
Physical chemistry chemical physics : PCCP, 8(1), 114-121 (2006-02-17)
Two-color (1 + 1') REMPI mass spectra of o-, m- and p-fluorophenol.ammonia (1 ration) clusters were measured with a long delay time between excitation and ionization lasers. The appearance of NH(4)(NH(3))(n-1)(+) with 100 ns delay after exciting the S(1) state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service