Skip to Content
Merck
All Photos(1)

Documents

105953

Sigma-Aldrich

2,4-Dichlorophenol

99%

Synonym(s):

1,3-Dichloro-4-hydroxybenzene, 2,4-DCP, 2,4-Dichlorophenic acid, 4,6-Dichlorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein:
742467
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050432
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

209-210 °C (lit.)

mp

42-43 °C (lit.)

solubility

methanol: soluble 1g in 10ml
ethanol: soluble 50 mg/mL

SMILES string

Oc1ccc(Cl)cc1Cl

InChI

1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H

InChI key

HFZWRUODUSTPEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dichlorophenol is anthropogenic organic compound and a common water and soil pollutant. It is precursor required for the synthesis of herbicides:2,4-dichloro- and 2,4,5-trichloro-phenoxyacetic acids and wood preservative: pentachlorophenol.

Application

2,4-Dichlorophenol was used to study the degradative pathway of 2,4-dichlorophenol by Phanerochaete chrysosporium. It was used to study the competitive sorption of anthropogenic organic compounds in soils.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

231.1 °F - closed cup

Flash Point(C)

110.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Valli et al.
Journal of bacteriology, 173(1), 345-352 (1991-01-01)
Under secondary metabolic conditions the white rot basidiomycete Phanerochaete chrysosporium mineralizes 2,4-dichlorophenol (I). The pathway for the degradation of 2,4-dichlorophenol (I) was elucidated by the characterization of fungal metabolites and of oxidation products generated by purified lignin peroxidase and manganese
Competitive sorption between 1, 3-dichlorobenzene or 2, 4-dichlorophenol and natural aromatic acids in soil organic matter.
Xing B and Pignatello JJ.
Environmental Science & Technology, 32(5), 614-619 (1998)
Efstratios Nikolaivits et al.
International journal of molecular sciences, 21(9) (2020-05-13)
2,4-Dichlorophenol (2,4-DCP) is a ubiquitous environmental pollutant categorized as a priority pollutant by the United States (US) Environmental Protection Agency, posing adverse health effects on humans and wildlife. Bioremediation is proposed as an eco-friendly, cost-effective alternative to traditional physicochemical remediation
Jiang Xu et al.
Journal of hazardous materials, 244-245, 628-636 (2012-11-28)
The Pd/FeFe(3)O(4) nanocomposites showed higher dechlorination efficiency of 2,4-dichlorophenol (2,4-DCP) rather than bare Pd/Fe nanoparticles in the batch dechlorination experiments. nFe(3)O(4) provided a convenient way to recycle the nanocomposites with an external magnetic field and significantly enhanced 2,4-DCP dechlorination. 2.0
Anwei Chen et al.
Chemosphere, 109, 208-212 (2014-02-18)
In this study, the H2S donor, sodium hydrosulfide (NaHS) was used to pretreat Phanerochaete chrysosporium in order to improve its ability to degrade 2,4-dichlorophenol (2,4-DCP). When pretreated with 100μM NaHS, P. chrysosporium was able to degrade 2,4-DCP completely in 24h

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service