Skip to Content
Merck
All Photos(1)

Key Documents

19422

Supelco

1-Butanol

analytical standard

Synonym(s):

n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
969148
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.55 (vs air)

vapor pressure

5 (4 mmHg) at 20 °C hPa

Assay

≥99.9% (GC)

autoignition temp.

649 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

11.2 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

refractive index

n20/D 1.399 (lit.)
n20/D 1.399

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butanol, alcohol with higher energy density, lower vapour pressure and low hygroscopicity. 1-Butanol is a potential fuel substitute to gasoline. It is also a feedstock used to produce chemicals. It is produced via two pathways, the synthetic 2-ketoacid pathway and CoA-dependent pathway. It can also be produced by fermentation of biomass-derived carbohydrates.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

It was used as a constituent in preparation of internal standard for analysis of fat-soluble vitamins using micellar electrokinetic chromatography (MEKC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Pedersen-Bjergaard et al.
Journal of chromatography. A, 876(1-2), 201-211 (2000-05-24)
Microemulsion electrokinetic chromatography (MEEKC) was carried out in a pH 2.5 phosphate buffer to effectively suppress the electroosmotic flow (EOF). With 66.6% (w/w) 25 mM phosphate buffer pH 2.5, 20.0% (w/w) 2-propanol, 6.6% (w/w) 1-butanol, 6.0% (w/w) sodium lauryl sulphate
Metabolic engineering of cyanobacteria for 1-butanol production from carbon dioxide.
Lan, Ethan I., and James C. Liao.
Metabolic engineering, 13.4, 353-363 (2011)
Production of dimethylfuran for liquid fuels from biomass-derived carbohydrates.
Roman-Leshkov, Yuriy, et al.
Nature, 447.7147, 982-985 (2007)
Metabolic engineering of< i> Escherichia coli</i> for 1-butanol and 1-propanol production via the keto-acid pathways.
Shen, Claire R., and James C. Liao.
Metabolic engineering, 10.6, 312-320 (2008)
Joel G Davis et al.
Nature, 491(7425), 582-585 (2012-11-23)
Hydrophobic hydration is considered to have a key role in biological processes ranging from membrane formation to protein folding and ligand binding. Historically, hydrophobic hydration shells were thought to resemble solid clathrate hydrates, with solutes surrounded by polyhedral cages composed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service