Merck
  • Extractionless method for the simultaneous high-performance liquid chromatographic determination of urinary caffeine metabolites for N-acetyltransferase 2, cytochrome P450 1A2 and xanthine oxidase activity assessment.
Extractionless method for the simultaneous high-performance liquid chromatographic determination of urinary caffeine metabolites for N-acetyltransferase 2, cytochrome P450 1A2 and xanthine oxidase activity assessment.
Journal of chromatography. B, Biomedical sciences and applications (2001-06-08)
A Nyéki, J Biollaz, U W Kesselring, L A Décosterd
ABSTRACT
Urinary metabolic ratios of caffeine are used in humans to assess the enzymatic activities of cytochrome P450 isoenzyme 1A2 (CYP1A2), xanthine oxidase (XO) and for phenotyping individuals for the bimodal N-acetyltransferase 2 (NAT2), all of them involved in the activation or detoxification of various xenobiotic compounds. Most reported analytical procedures for the measurement of the urinary metabolites of caffeine include a liquid-liquid extraction of urine samples prior to their analysis by reversed-phase HPLC. At neutral to basic pH however, 5-acetylamino-6-formylamino-3-methyluracil (AFMU), a metabolite of caffeine, spontaneously decomposes to 5-acetylamino-6-amino-3-methyluracil (AAMU). Since AAMU is not extracted in most organic solvents, the extent of AFMU decomposition cannot be precisely assessed. Although the decomposition reaction can be minimized by immediate acidification of the urine, accurate results can only be obtained when both AAMU and AFMU are monitored, or alternatively, if AAMU is measured after complete transformation of AFMU into AAMU in basic conditions. We report a liquid chromatographic method for the simultaneous quantitative analysis of the five urinary metabolites of caffeine used for the CYP1A2, XO and NAT2 phenotyping studies: AAMU, AFMU, 1-methylxanthine, 1-methyluric acid and 1,7-dimethyluric acid. These metabolites are satisfactory separated from all other known caffeine metabolites as well as endogenous urinary constituents. Sample treatment does not require any liquid-liquid extraction procedure. Urine samples are diluted and centrifuged before being injected (10 microl) onto a YMC-Pack Polyamine II (250x4.6 mm) column. A step-wise gradient elution program is applied using acetonitrile-0.75% (v/v) formic acid: (91:9) at 0 min-->(75:25) at 25 min-->(65:35) at 35 min-->(65:35) at 45 min, followed by a re-equilibration step to the initial solvent composition. The flow-rate is 1.0 ml/min and the separations are monitored by UV absorbance at 260 and 280 nm. The procedure described here represents a substantial improvement over previous methods: a single analysis and a minimal urine sample treatment enables the simultaneous quantitation of five caffeine metabolites, notably AFMU and AAMU, used for the determination of CYP450 1A2, XO and NAT2 enzyme activity. Importantly enough, phenotyping individuals for the bimodal NAT2 is made possible without the uncertainty associated with the deformylation of AFMU, which is likely to happen at all steps prior to the analysis, during sample storage and even in the bladder of the subjects.
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
SAFC
L-Valine
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Reagent Alcohol, reagent grade
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Hexanal, ≥97%, FCC, FG
Sigma-Aldrich
Hexanal, 98%