Skip to Content
Merck
All Photos(1)

Key Documents

94619

Sigma-Aldrich

L-Valine

≥99.5% (NT), BioUltra

Synonym(s):

(S)-α-Aminoisovaleric acid, L-2-Amino-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH(NH2)CO2H
CAS Number:
Molecular Weight:
117.15
Beilstein:
1721136
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

L-Valine, BioUltra, ≥99.5% (NT)

product line

BioUltra

Assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D +27.0±0.5°, c = 5% in 5 M HCl

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.05%

loss

≤0.05% loss on drying, 20 °C (HV)

color

white

mp

295-300 °C (subl.) (lit.)

solubility

1 M HCl: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤150 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.5 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 0.15
λ: 280 nm Amax: 0.1

SMILES string

CC(C)[C@H](N)C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1

InChI key

KZSNJWFQEVHDMF-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Metabolomics reveals metabolites associated with hair follicle cycle in cashmere goats.: This study utilizes L-Valine as a standard for detecting, understanding, and enhancing hair follicle cycles in cashmere goats through metabolomic analysis, aiding in the improvement of fiber production and quality (Ma et al., 2024).

Other Notes

Protects enzymes against heat inactivation[1]; As spacer in isotachophoresis of serum proteins[2][3]; Growth requirement of various microorganisms[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Yagi et al. et al.
Electrophoresis ?83, 503-503 (1984)
CRC Handbook of Microbiology, 4, 1-1 (1974)
L.G. Paleg et al.
Australian Journal of Plant Physiology, 8, 107-107 (1981)
T. Hine
Electrophoresis ?83, 541-541 null
Shankar Sachidhanandam et al.
Nature neuroscience, 16(11), 1671-1677 (2013-10-08)
Neocortical activity can evoke sensory percepts, but the cellular mechanisms remain poorly understood. We trained mice to detect single brief whisker stimuli and report perceived stimuli by licking to obtain a reward. Pharmacological inactivation and optogenetic stimulation demonstrated a causal

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service