Skip to Content
Merck
All Photos(3)

Key Documents

277649

Sigma-Aldrich

Reagent Alcohol

anhydrous reagent, contains 5% isopropyl alcohol and 5% methyl alcohol as denaturant

Synonym(s):

Ethyl alcohol, Reagent alcohol, Ethanol, RGA, denatured alcohol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
reagent grade
bp:
78 °C

Product Name

Reagent Alcohol, anhydrous, ≤0.003% water

grade

anhydrous
reagent grade

Quality Level

vapor density

1.59 (vs air)

form

liquid

contains

5% isopropyl alcohol as denaturant
5% methyl alcohol as denaturant

expl. lim.

3.1-27.7 %

concentration

100%

impurities

≤0.003% water
≤0.005% water (100 mL pkg)

evapn. residue

<0.0005%

bp

78 °C

mp

-114 °C

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reagent Alcohol is ethanol that is made non-consumable by the addition of denaturants.

Components

95 parts of specially denatured ethyl alcohol 3A, 200 proof, with 5 parts of isopropyl alcohol. Final concentrations are ~ 90% ethanol, ~ 5% methanol and ~ 5% isopropanol.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surrogate alcohol: what do we know and where do we go?.
Lachenmeier DW, et al
Alcoholism, Clinical and Experimental Research, 31(10), 1613-1624 (2007)
Effects of added dibutyl phosphate on the luminescent properties of europium tetrakis dibenzoylmethide triethylammonium.
Fontenot RS, et al.
Journal of Luminescence, 158, 428-434 (2015)
Metabolite and Fatty Acid Analysis of Yeast Cells and Culture Supernatants.
Chen L and Chen WN.
Bio-protocol, 4(17) (2014)
Surrogate alcohol: what do we know and where do we go?
Lachenmeier DW, et al.
Alcoholism, Clinical and Experimental Research, 31(10), 1613-1624 (2007)
Eagleson M.
Concise Encyclopedia Chemistry, 384-384 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service