Skip to Content
Merck
All Photos(1)

Documents

E-035

Supelco

Ethanol-25

25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®

Synonym(s):

Ethanol solution, Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 10 × 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

25 mg/dL in H2O

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A kit of NIST traceable ethanol standards suitable for calibration of ethanol testing methods. These Certified Reference Materials are widely used in forensic analysis and clinical toxicology applications for determination of blood alcohol content.

Application


  • Exopolysaccharide purification: Raj et al. utilized ethanol as part of an alcohol/salt-based aqueous two-phase system to purify exopolysaccharides from Lactobacillus spp., demonstrating its effectiveness in biotechnological applications and showcasing ethanol′s role in enhancing antidiabetic properties of biomolecules (Raj et al., 2024).

  • Antioxidant phenolic compounds extraction: De Jesus et al. conducted a study using ethanol for the microwave and ultrasound-assisted extraction of antioxidant phenolic compounds from Lantana camara leaves, highlighting ethanol′s utility in optimizing extraction processes for natural product research and phytochemical analysis (de Jesus et al., 2024).

  • Biopolymer degradation and ester production: Malunavicius et al. reported on the use of ethanol in the enzymatic degradation of polycaprolactones and simultaneous ester production, emphasizing ethanol′s role in sustainable biopolymer research and industrial applications (Malunavicius et al., 2023).

  • Coffee silverskin properties analysis: Buyong and Nillian explored the physiochemical properties of Liberica coffee silverskin using various solvents including ethanol, underscoring ethanol′s importance in food science and nutritional studies for extracting valuable compounds from agricultural by-products (Buyong and Nillian, 2023).

  • Polyphenol extraction from Acanthopanax senticosus: Li et al. employed ethanol in an ionic liquid-assisted aqueous two-phase system for the extraction and purification of polyphenols from Acanthopanax senticosus, demonstrating ethanol′s applicability in enhancing the efficiency of bioactive compound isolation in pharmaceutical research (Li et al., 2023).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Enocksson et al.
The Journal of clinical investigation, 95(5), 2239-2245 (1995-05-01)
Although it is well established in several mammalian species that beta 3-adrenoceptors play a major role in regulating lipolysis and thermogenesis in adipose tissue, the functional existence and role of this receptor subtype in man has been controversial. We investigated
Thiago M Pais et al.
PLoS genetics, 9(6), e1003548-e1003548 (2013-06-12)
The yeast Saccharomyces cerevisiae is able to accumulate ≥17% ethanol (v/v) by fermentation in the absence of cell proliferation. The genetic basis of this unique capacity is unknown. Up to now, all research has focused on tolerance of yeast cell
A Nyéki et al.
Journal of chromatography. B, Biomedical sciences and applications, 755(1-2), 73-84 (2001-06-08)
Urinary metabolic ratios of caffeine are used in humans to assess the enzymatic activities of cytochrome P450 isoenzyme 1A2 (CYP1A2), xanthine oxidase (XO) and for phenotyping individuals for the bimodal N-acetyltransferase 2 (NAT2), all of them involved in the activation
William Chan et al.
The Canadian journal of cardiology, 30(8), 912-919 (2014-07-30)
Myocardial contrast echocardiography during angiography is critical in identifying appropriate septal perforator(s) for alcohol septal ablation (ASA) in patients with hypertrophic obstructive cardiomyopathy. We evaluated whether there were other angiographic and/or echocardiographic markers that might identify patients who are anatomically
Irene Zelner et al.
Drug metabolism reviews, 45(3), 277-299 (2013-05-30)
The ability to undergo non-oxidative metabolism from ethanol to fatty acid ethyl esters (FAEEs) varies greatly among tissues and organs. To gain a greater understanding of non-oxidative ethanol metabolism to FAEE, we aimed to collect all published data on FAEE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service