Merck
  • Neural computational prediction of oral drug absorption based on CODES 2D descriptors.
Neural computational prediction of oral drug absorption based on CODES 2D descriptors.
European journal of medicinal chemistry (2009-12-22)
A Guerra, N E Campillo, J A Páez
ABSTRACT
A neural model based on a numerical molecular representation using CODES program to predict oral absorption of any structure is described. This model predicts both high and low-absorbed compounds with a global accuracy level of 74%. CODES/ANN methodology shows promising utilities not only as a conventional in silico tool in high-throughput screening or improvement of absorption capabilities procedures but also the improvement of in vitro-in vivo correlation could be addressed.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Chloroform solution, NMR reference standard, 50% in acetone-d6 (99.9 atom % D), chromium(III) acetylacetonate 0.2 %
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Chloroform solution, NMR reference standard, 3% in acetone-d6 (99.9 atom % D)
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Chloroform solution, certified reference material, 5000 μg/mL in methanol
Supelco
Nicotinic acid, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Oxazepam
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Prednisolone, ≥99%