Skip to Content
Merck
All Photos(5)

Documents

N4126

Sigma-Aldrich

Nicotinic acid

≥98%

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥98%

form

powder

color

white to off-white

mp

236-239 °C (lit.)

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)
rat ... Chrnb2(54239)

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic acid is also known as vitamin B3 or Niacin. It is a water soluble vitamin. Niacin can be either synthesized within the body or can be obtained from dietary sources.

Application

Nicotinic acid has been used:
  • as a component of the MS vitamins stock solution
  • for the detection of siderophores in Escherichia coli bacterial culture
  • as a component of the holidic medium for adult Drosophila .

Biochem/physiol Actions

Niacin is involved in the production of nicotinamide adenine dinucleotide (NAD+), which is used in the adenine triphosphate (ATP) production in mitochondrial oxidative phosphorylation. It also decreases hepatic production of very-low density lipoproteins (VLDL), and apolipoprotein B (apo-B). Niacin deficiency leads to pellagra.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin B3 and sirtuin function
Denu J M
Trends in Biochemical Sciences, 30(9), 479-483 (2005)
Mechanism of action of niacin
Kamanna V S and Kashyap M L
The American Journal of Cardiology, 101(8), S20-S26 (2008)
A holidic medium for Drosophila melanogaster
Piper M D W, et al.
Nature Methods, 11(1), 100-100 (2014)
A procedure allowing up to eight generations of wheat and nine generations of barley per annum
Zheng Z, et al.
Euphytica, 191(2), 311-316 (2013)
An investigation into the pathogenic properties of Escherichia coli strains BLR, BL21, DH5 alpha and EQ1
Chart H, et al.
Journal of Applied Microbiology, 89(6), 1048-1058 (2000)

Articles

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Lipoproteins package cholesterol for transport in plasma, essential for lipid transport and cellular function in the body.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service