Skip to Content
Merck
All Photos(2)

Key Documents

A3160

Sigma-Aldrich

Acetylsalicylic acid

analytical standard

Synonym(s):

2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
779271
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

analytical standard

form

powder

packaging

vial of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

134-136 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical
veterinary

SMILES string

CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

BSYNRYMUTXBXSQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

Packaging

Supplied in amber screw-cap vials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

482.0 °F

Flash Point(C)

250 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niels Hulsman et al.
Journal of medicinal chemistry, 50(10), 2424-2431 (2007-04-20)
Hybrid drug 1 (NO-ASA) continues to attract intense research from chemists and biologists alike. It consists of ASA and a -ONO2 group connected through a spacer and is in preclinical development as an antitumor drug. We report that, contrary to
Connie N Hess et al.
Journal of the American College of Cardiology, 66(7), 777-787 (2015-08-14)
Bleeding limits anticoagulant treatment in patients with acute coronary syndromes (ACS). We investigated whether background concomitant antiplatelet therapy influences the effects of apixaban after ACS. This study examined high-risk ACS patients who were treated with aspirin or aspirin plus clopidogrel
Andrew O Maree et al.
Journal of the American College of Cardiology, 46(7), 1258-1263 (2005-10-04)
We investigated whether use of low-dose enteric-coated (EC) aspirin for secondary prevention of cardiovascular events has sufficient bioavailability to achieve complete platelet cyclooxygenase (COX) inhibition in all individuals. Aspirin reduces cardiovascular morbidity and mortality in patients with pre-existing vascular disease;
P J Devereaux et al.
The New England journal of medicine, 370(16), 1494-1503 (2014-04-01)
There is substantial variability in the perioperative administration of aspirin in patients undergoing noncardiac surgery, both among patients who are already on an aspirin regimen and among those who are not. Using a 2-by-2 factorial trial design, we randomly assigned
Reiko Nishihara et al.
JAMA, 309(24), 2563-2571 (2013-06-27)
Aspirin use reduces the risk of colorectal carcinoma. Experimental evidence implicates a role of RAF kinases in up-regulation of prostaglandin-endoperoxide synthase 2 (PTGS2, cyclooxygenase 2), suggesting that BRAF-mutant colonic cells might be less sensitive to the antitumor effects of aspirin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service