Skip to Content
Merck
Water suitable for UV/Vis spectroscopy

Water

Water suitable for UV/Vis spectroscopy
Synonym(s):
Water
Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Sterilization Method
Technique
Showing 1-20 of 39
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2