Skip to Content
Merck
All Photos(2)

Key Documents

38796

Sigma-Aldrich

Water

Deionized

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2O
CAS Number:
Molecular Weight:
18.02
Beilstein:
2050024
EC Number:
MDL number:
UNSPSC Code:
12191602
PubChem Substance ID:
NACRES:
NA.21
bp:
100 °C (lit.)
vapor pressure:
3 mmHg

vapor density

<1 (vs air)

vapor pressure

3 mmHg

form

liquid

quality

Deionized

dilution

(for analytical testing)

refractive index

n20/D 1.34 (lit.)

pH

6.0-8.0 (25 °C)

conductivity

≤4.3 μS/cm

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

SMILES string

O

InChI

1S/H2O/h1H2

InChI key

XLYOFNOQVPJJNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Water (deionized) is a clear colorless liquid having boiling point 100°C (lit) and refractive index n20/D 1.34(lit.).

Application

Water (deionized) may be used for the following studies:
  • Rinsing of slides.
  • For the elimination of surfactant and excess silver ions during the preparation of silver nanoparticles.
  • Preparation of powdered milk formula samples for the quantification of fluoride.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A simple technique for demonstrating centromeric heterochromatin.
A T Sumner
Experimental cell research, 75(1), 304-306 (1972-11-01)
M A Buzalaf et al.
ASDC journal of dentistry for children, 68(1), 37-41 (2001-04-28)
Usually infant milk formula is the major source of fluoride in infancy. Fluoride concentrations in ten samples of powdered milk formulas, prepared with deionized, bottled mineral, and fluoridated drinking water were determined after HMDS-facilitated diffusion, using a fluoride ion specific
Ivan Sondi et al.
Journal of colloid and interface science, 275(1), 177-182 (2004-05-26)
The antimicrobial activity of silver nanoparticles against E. coli was investigated as a model for Gram-negative bacteria. Bacteriological tests were performed in Luria-Bertani (LB) medium on solid agar plates and in liquid systems supplemented with different concentrations of nanosized silver
Susu Duan et al.
Nature communications, 5, 5029-5029 (2014-10-10)
Oseltamivir-resistant H1N1 influenza viruses carrying the H275Y neuraminidase mutation predominated worldwide during the 2007-2009 seasons. Although several neuraminidase substitutions were found to be necessary to counteract the adverse effects of H275Y, the order and impact of evolutionary events involved remain
Mary A Rodgers et al.
The Journal of experimental medicine, 211(7), 1333-1347 (2014-06-25)
Linear ubiquitination is a newly discovered posttranslational modification that is currently restricted to a small number of known protein substrates. The linear ubiquitination assembly complex (LUBAC), consisting of HOIL-1L, HOIP, and Sharpin, has been reported to activate NF-κB-mediated transcription in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service