great purity for LCMS with use with Absciex for toxicology panel
WX0001
Water
suitable for LC/MS, OmniSolv®, suitable for SM 4500 - NH3
Synonym(s):
Aqua, Dihydrogen oxide
About This Item
LC/MS: suitable
Recommended Products
Product Name
Water, Suitable for LCMS chromatography, OmniSolv® (Updated non-compliant text)
Agency
suitable for SM 4500 - NH3
Quality Level
vapor pressure
23 hPa ( 20 °C)
product line
OmniSolv®
form
liquid
shelf life
12 mo. (from manufacture date)
technique(s)
HPLC: suitable
LC/MS: suitable
impurities
≤50 ppb p-nitrophenol (LC/MS)
≤50 ppb reserpine (LC/MS)
evapn. residue
≤1 ppm
refractive index
n20/D 1.34 (lit.)
pH
6.0- 8.0 (25 °C, 77 °F)
bp
100 °C (lit.)
mp
0 °C (lit.)
solubility
water: miscible
density
1.000 g/mL at 3.98 °C (lit.)
UV cutoff
≤190 nm
UV absorption
λ: 200 nm Amax: ≤0.01
λ: 210 nm Amax: ≤0.01
λ: 220 nm Amax: ≤0.01
λ: 230 nm Amax: ≤0.01
λ: 240 nm Amax: ≤0.01
λ: 250 nm Amax: ≤0.005
λ: 270 nm Amax: ≤0.005
λ: 400-280 nm Amax: ≤0.005
application(s)
general analytical
shipped in
ambient
storage temp.
room temp
SMILES string
O
InChI
1S/H2O/h1H2
InChI key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Preparation Note
Analysis Note
Evaporation residue: 1ppm max
Filtered through 0.2 m filter: To pass test
Fluorescence as quinine at 254 nm: 1ppb max
Fluorescence as quinine at 365 nm: 1ppb max
Gradient at 210 nm: 2mAU max
Gradient at 254 nm: 0.5mAU max
LC/MS Purity (p-nitrophenol): 50ppb max
LC/MS Purity (reserpine): 50ppb max
Metals, suitable for LC/MS: Passes test
UV Abs. at 200 nm: 0.01AU max
UV Abs. at 210 nm: 0.01AU max
UV Abs. at 220 nm: 0.01AU max
UV Abs. at 230 nm: 0.01AU max
UV Abs. at 240 nm: 0.01AU max
UV Abs. at 250 nm: 0.005AU max
UV Abs. at 270 nm: 0.005AU max
UV Abs. at 400-280 nm: 0.005AU max
UV Cut-off: 190nm max
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
In this article we provide a complete workflow of quantification and identification of acylcarnitines in dried blood spots (DBS) using stable isotope-labeled (SIL) acylcarnitines CRMs by FIA-MS/MS.
Related Content
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service