Skip to Content
Merck
All Photos(1)

Key Documents

1269618

USP

Fenofibrate Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, Fenofibric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO4
CAS Number:
Molecular Weight:
318.75
Beilstein:
2058973
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

fenofibrate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C(C1=CC=C(OC(C)(C)C(O)=O)C=C1)C2=CC=C(Cl)C=C2

InChI

1S/C17H15ClO4/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10H,1-2H3,(H,20,21)

InChI key

MQOBSOSZFYZQOK-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fenofibrate Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fenofibrate
  • Fenofibrate Capsules
  • Fenofibrate Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roger K Verbeeck et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 18(1), 61-67 (2015-04-17)
The therapeutic equivalence of multiple registered fenofibrate formulations, several of which are suprabioavailable and therefore marketed at lower dosage strengths than their reference products, is based on the results of bioequivalence studies. Most of these formulations show a higher bioavailability
Yunxia Lu et al.
Biochemical and biophysical research communications, 458(1), 1-7 (2015-01-17)
Fenofibrate (FF) is widely used to lower blood lipids in clinical practice, but whether its protective effect on endothelium-dependent vasodilatation (EDV) in thoracic aorta is related with endoplasmic reticulum (ER) stress remains unknown. In this study, female Sprauge Dawley rats
Martin J A Schackmann et al.
Biochimica et biophysica acta, 1851(2), 231-237 (2014-12-17)
X-linked adrenoleukodystrophy (X-ALD) is a neurometabolic disease that is caused by mutations in the ABCD1 gene. ABCD1 protein deficiency impairs peroxisomal very long-chain fatty acid (VLCFA) degradation resulting in increased cytosolic VLCFA-CoA levels, which are further elongated by the VLCFA-specific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service