Skip to Content
Merck
All Photos(1)

Key Documents

F0048010

Fenofibrate impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, Fenofibric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO4
CAS Number:
Molecular Weight:
318.75
Beilstein:
2058973
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fenofibrate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(OC(C)(C)C(O)=O)C=C1)C2=CC=C(Cl)C=C2

InChI

1S/C17H15ClO4/c1-17(2,16(20)21)22-14-9-5-12(6-10-14)15(19)11-3-7-13(18)8-4-11/h3-10H,1-2H3,(H,20,21)

InChI key

MQOBSOSZFYZQOK-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fenofibrate impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Aldini et al.
British journal of pharmacology, 74(3), 611-617 (1981-11-01)
1 Hypolipidaemic agents may increase biliary cholesterol in man, inducing a supersaturated bile. 2 To evaluate this possible side-effect, we have studied bile lipid secretion over a period of 8 h with intact enterohepatic circulation and 4 h with complete
Simple and rapid method for determining procetofenic acid, an active metabolite of procetofen, in biological fluids by solid-phase extraction and high-performance liquid chromatography.
A C Ramusino et al.
Journal of chromatography, 383(2), 419-424 (1986-12-19)
Inactivation of 3-hydroxy-3-methylglutaryl coenzyme A reductase in cultured human blood mononuclear cells by procetofenic acid.
C Massaroli et al.
Pharmacological research communications, 14(1), 51-60 (1982-01-01)
Rapid determination of procetofenic acid in plasma by high-performance liquid chromatography.
M C Caturla et al.
Journal of chromatography, 227(1), 219-222 (1982-01-08)
S Pourbaix et al.
Biochemical pharmacology, 33(22), 3661-3666 (1984-11-15)
Hamsters were given a diet containing fenofibrate (0.5% or 0.05%) or its metabolite, LF 2151 (0.15% or 0.015%) or a standard diet for a 3-week period. At the end of this period, the analysis of plasma lipids showed that the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service