Skip to Content
Merck
All Photos(1)

Documents

40062

Supelco

Pentachlorophenol solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1285380
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6HCl5O/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H

InChI key

IZUPBVBPLAPZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup - Solvent

Flash Point(C)

9.7 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junfeng Niu et al.
Journal of hazardous materials, 246-247, 119-125 (2013-01-09)
Horseradish peroxidase (HRP) is successfully in situ encapsulated into the poly(D,L-lactide-co-glycolide) (PLGA)/PEO-PPO-PEO (F108) electrospun fibrous membranes (EFMs) by emulsion electrospinning. The adsorption and degradation of pentachlorophenol (PCP) by HRP-EFMs are investigated. The experimental results show that the sorption kinetic of
Lee Karlsson et al.
American journal of public health, 103(2), 276-277 (2012-12-15)
In 2009, after resident calls regarding an odor, the Vermont Department of Health and state partners responded to 2 scenarios of private drinking water contamination from utility poles treated with pentachlorophenol (PCP), an organochlorine wood preservative used in the United
Jiajiang Lin et al.
Environmental pollution (Barking, Essex : 1987), 178, 433-440 (2013-05-01)
Pentachlorophenol (PCP) dissipation occurs naturally in flooded soils and although dissipation half-lives vary between soil profiles at the millimeter-scale the reason is poorly understood. Vertical variations of PCP dissipation were investigated in three typical Chinese paddy soils; Soil 1 (Umbraqualf)
Min Yang et al.
Journal of hazardous materials, 237-238, 63-70 (2012-09-12)
In this study, an imprinted silica matrix of pentachlorophenol (PCP) co-loaded with Fe(3)O(4) nanoparticles and ZnS:Mn(2+) quantum dots (QDs) was fabricated. The introduction of Fe(3)O(4) nanoparticles to the imprinted matrix provided an easy way to separate PCP under an external
Tasia Hurd et al.
Journal of applied toxicology : JAT, 32(8), 627-634 (2012-08-23)
Pentachlorophenol (PCP) is an organochlorine pesticide that decreases the tumor-cell killing (lytic) function of human natural killer (NK) cells. NK cells defend against tumor cells and virally infected cells. They bind to these targets, utilizing a variety of cell-surface proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service