Skip to Content
Merck
All Photos(1)

Documents

442284

Supelco

2,3,5,6-Tetrachlorophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl4O
CAS Number:
Molecular Weight:
231.89
Beilstein:
2049586
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Oc1c(Cl)c(Cl)cc(Cl)c1Cl

InChI

1S/C6H2Cl4O/c7-2-1-3(8)5(10)6(11)4(2)9/h1,11H

InChI key

KEWNKZNZRIAIAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,5,6-Tetrachlorophenol belongs to the class of substituted phenol compounds, which are widely used in industrial applications which include synthesis of pesticides, dyes, drugs, plastics, etc. They are found in environmental samples as pollutant due to their extensive usage.

Application

2,3,5,6-Tetrachlorophenol may be used as an analytical reference standard for the determination of the analyte in landfill leachates, aqueous samples, and environmental samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Chauhan et al.
Applied microbiology and biotechnology, 52(2), 261-266 (1999-09-28)
Degradation of 2,4,5-trichlorophenol (2,4,5-TCP) and 2,3,5,6-tetrachlorophenol (TeCP) was studied using a two-stage approach that utilized efficient pulse electric discharge (PED) followed by biological degradation with a consortium from acclimated return activated sludge. The chlorinated phenols were treated in the PED
Evaluation of a dedicated gas chromatography-mass spectrometry method for the analysis of phenols in water.
Vermeulen A, et al.
Journal of Chromatography A, 1071(1-2), 41-46 (2005)
Acute dermal toxicity of tetrachlorophenols in the rat.
S Y Shen et al.
Bulletin of environmental contamination and toxicology, 31(6), 680-685 (1983-12-01)
Development and optimization of a method for the analysis of phenols and chlorophenols from aqueous samples by gas chromatography-mass spectrometry, after solid-phase extraction and trimethylsilylation.
Kovacs A, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 99(1), 125-131 (2011)
Determination of chlorophenols by solid-phase microextraction and liquid chromatography with electrochemical detection.
Sarrion NM, et al.
Journal of Chromatography A, 947(2), 155-165 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service