Skip to Content
Merck
All Photos(1)

Key Documents

D201154

Sigma-Aldrich

Dioctyl phthalate

≥99.5%

Synonym(s):

Bis(2-ethylhexyl) phthalate, DEHP, DOP, Phthalic acid bis(2-ethylhexyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
390.56
Beilstein:
1890696
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>16 (vs air)

Quality Level

vapor pressure

1.2 mmHg ( 93 °C)

Assay

≥99.5%

form

oil

autoignition temp.

734 °F

impurities

≤0.05% water (Karl Fischer)

color

APHA: ≤10

refractive index

n25/D 1.483-1.487
n20/D 1.486 (lit.)

bp

384 °C (lit.)

mp

−50 °C (lit.)

density

0.985-0.987 g/mL at 20 °C
0.985 g/mL at 25 °C (lit.)

suitability

suitable for acidity (

<=0.003%as phthalic acid

)

SMILES string

CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dioctyl phthalate is a non-volatile solvent mainly used as a plasticizer for polymers such as polyvinyl chloride (PVC), polystyrene (PS) and polyisoprene (PI).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic mechanical behavior of the dioctyl phthalate plasticized polyvinyl chloride?epoxidized soya bean oil.
Semsarzadeh MA
European Polymer Journal, 38(2), 351-358 (2002)
Shear effects on solvated block copolymer lamellae: Polystyrene-polyisoprene in dioctyl phthalate.
Balsara NP
Physical Review Letters, 72(3), 360-360 (1994)
Zhi-Ping Du et al.
Toxicology and applied pharmacology, 394, 114960-114960 (2020-03-24)
During pregnancy, fetal thyroid hormones (THs) are dependent on maternal placental transport and their physiological level is crucial for normal fetal neurodevelopment. Earlier research has shown that Di-(2-ethylhexyl) phthalate (DEHP) disrupts thyroid function and THs homeostasis in pregnant women and
Minjeong Kim et al.
Biomolecules & therapeutics, 26(5), 512-519 (2018-08-07)
Phthalates widely used in the manufacture of plastics have deeply penetrated into our everyday lives. Recently, a concern over the toxicity of phthalates on thyroid, has been raised but in most of cases, the doses employed were unrealistically high. To
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 43(34), 13122-13135 (2014-07-22)
Eighteen Mn complexes with N-donor and carboxylate ligands have been synthesized and characterized. Three Mn complexes among them are new and are reported for the first time. The reactions of oxygen evolution in the presence of oxone (2KHSO5·KHSO4·K2SO4) and cerium(iv)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service