Skip to Content
Merck
All Photos(2)

Key Documents

T9179

Sigma-Aldrich

Terrific Broth

EZMix powder microbial growth medium

Synonym(s):

TB media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

for molecular biology

Quality Level

description

TB; quick dissolve, dust-free formulation

sterility

non-sterile

form

powder

technique(s)

microbiological culture: suitable

application(s)

agriculture
food and beverages
microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Terrific Broth is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains glycerol, peptides, amino acids, water-soluble vitamins, and carbohydrates.

Application

Suitable for non-selective cultivation of E. coli strains for cloning, high yield DNA plasmid production and high yield production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added.
Terrific broth has been used:
  • as a medium for selective growth of E.coli BL21 (DE3) V2RpRARE expressing MLL SET (nuclear proto-oncogene) domain in pET28GST-LIC
  • to grow BL21 (DE3) cells containing plasmid expressing IL-8 (interleukin-8)
  • Terrific broth is also suitable for use in bacterial suspension (Bacillus subtilis strain 1085B)

Features and Benefits

Terrific Broth EZMix powder provides:
  • Granulated, dust-free format for safer handling and faster mixing
  • A budget-friendly alternative to liquid
  • Modified formulation (without glycerol)
  • Easy scale-up using larger package sizes

Components

*Note some of the product sizes available are given in liquid measure. This is a powder format product and those sizes reflect the final reconstituted volume.

23.6g/L Yeast Extract
11.8g/L Tryptone
9.4g/L K2HPO4
2.2g/L KH2PO4

Reconstitution

Suspend 24.1 g of powder and 4 mL of glycerol in sufficient deionized water to make 500 mL. Autoclave for 15 minutes at 121 °C to sterilize. Allow to cool before making additions, such as antibiotics (if desired).

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul Peluso et al.
Analytical biochemistry, 312(2), 113-124 (2003-01-18)
Antibody microarrays have the potential to revolutionize protein expression profiling. The intensity of specific signal produced on a feature of such an array is related to the amount of analyte that is captured from the biological mixture by the immobilized
Idan Tuval et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(7), 2277-2282 (2005-02-09)
Aerobic bacteria often live in thin fluid layers near solid-air-water contact lines, in which the biology of chemotaxis, metabolism, and cell-cell signaling is intimately connected to the physics of buoyancy, diffusion, and mixing. Using the geometry of a sessile drop
Eve Toulza et al.
Applied and environmental microbiology, 76(13), 4521-4529 (2010-05-04)
Understanding the conditions leading to harmful algal blooms, especially those produced by toxic dinoflagellate species, is important for environmental and health safety. In addition to investigations into the environmental conditions necessary for the formation of toxic blooms, we postulate that
Optimizing antibody immobilization strategies for the construction of protein microarrays
Peluso P, et al.
Analytical Biochemistry, 312(2), 113-124 (2003)
Lac V Lee et al.
Biochemistry, 49(25), 5366-5376 (2010-05-19)
Undecaprenyl pyrophosphate synthase (UPPS) catalyzes the consecutive condensation of eight molecules of isopentenyl pyrophosphate (IPP) with farnesyl pyrophosphate (FPP) to generate the C(55) undecaprenyl pyrophosphate (UPP). It has been demonstrated that tetramic acids (TAs) are selective and potent inhibitors of

Protocols

General protocols for growth of competent cells in microbial medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service