Skip to Content
Merck
All Photos(2)

Key Documents

L3022

Sigma-Aldrich

LB Broth (Lennox)

Highly-referenced microbial growth powder medium, low salt, suitable for salt-sensitive E.coli culture.

Synonym(s):

Lennox broth, Lennox broth , Luria Bertani Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

grade

for molecular biology

sterility

non-sterile

form

powder

composition

NaCl, 5 g/L
Tryptone, 10 g/L
Yeast Extract, 5 g/L

technique(s)

microbiological culture: suitable

pH

6.8-7.2(2% solution)

application(s)

food and beverages
microbiology

storage temp.

room temp

suitability

nonselective for Escherichia coli
nonselective for coliforms

General description

Lennox LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids and carbohydrates in a low-salt formulation.

Application

Luria Broth is used for maintenance and propagation of Escherichia coli. The E. coli grow faster in Luria Broth because the tryptone and yeast supply essential growth factors that the E. coli would otherwise have to synthesize. Luria Broth also contains essential electrolytes for transport and osmotic balance, due to the NaCl component.
It was used for resuspension of T. pyriformis cell fractions in a study of Trimastix fractionation of cellular extracts.
Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added, including those that require low-salt conditions, such as Zeocin®.

Features and Benefits

Lennox LB powder provides:
  • Convenient small package to eliminate weighing
  • Easy scale-up using larger package sizes
  • A budget-friendly alternative to liquid
  • Standard formulation

Preparation Note

1. Suspend 20 g in 1 L of distilled water.
2. Autoclave for 15 minutes at 121 °C.
To prepare Lennox L Broth: Add 1 g glucose and proceed with preparation instructions as above.
To prepare the medium of Enquist and Sternberg: Aseptically add 10 ml of sterile 1 M magnesium sulfate after autoclaving.

Legal Information

Zeocin is a registered trademark of Cayla Sarl

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Benavente et al.
International journal of biological macromolecules, 103, 758-763 (2017-05-27)
The preparation of silver nanoparticles (AgNPs) and their incorporation into the structure of a regenerated cellulose membrane by dip coating is presented. Morphological characterization of the AgNPs (average diameter of 20±2nm) was carried out by SEM/TEM, while elastic, electrical and
Margot Doberva et al.
Frontiers in microbiology, 8, 1152-1152 (2017-07-12)
Quorum sensing (QS) is a density-dependent mechanism allowing bacteria to synchronize their physiological activities, mediated by a wide range of signaling molecules including
Francisco Dávila et al.
The Journal of experimental biology, 221(Pt 6) (2018-02-16)
Bacteria can damage sperm and thus reduce the reproductive success of both males and females; selection should therefore favour the evolution of antimicrobial protection. Eusocial hymenopterans might be particularly affected by such bacterial infections because of their mating ecology. In
I M Ogunade et al.
Journal of dairy science, 100(3), 1780-1794 (2017-01-04)
Inhibiting the growth of Escherichia coli O157:H7 (EC) in feeds may prevent the transmission or cycling of the pathogen on farms. The first objective of this study was to examine if addition of propionic acid or microbial inoculants would inhibit
Víctor González-Alonso et al.
Poultry science, 99(1), 536-545 (2020-05-18)
The objective of the present study was to assess the potential synergistic effect between supercritical carbon dioxide (SC-CO2) and fresh culinary herbs (Coriandrum sativum and Rosmarinus officinalis) on the microbial inactivation of raw chicken meat. The microbiological inactivation was performed

Protocols

General protocols for growth of competent cells in microbial medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service