Skip to Content
Merck
All Photos(2)

Key Documents

G5516

Sigma-Aldrich

Glycerol

for molecular biology, ≥99.0%

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
50161901
PubChem Substance ID:
NACRES:
NA.31
grade:
for molecular biology
reagent
Assay:
≥99.0%
technique(s):
MALDI-MS: suitable
cryopreservation: suitable
application(s):
life science and biopharma
microbiology
sample preservation
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)

grade

for molecular biology
reagent

Quality Level

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥99.0%

form

viscous liquid

autoignition temp.

698 °F

expl. lim.

≥2.7-19.0 %

technique(s)

MALDI-MS: suitable
cryopreservation: suitable

impurities

<5 ppm Iron
<5 ppm heavy metal
<5 ppm magnesium

refractive index

n20/D 1.474 (lit.)

pH

5.5-8

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

solubility

water: 1 mL/mL, clear, colorless

density

1.25 g/mL (lit.)

cation traces

Fe: ≤5 ppm
Mg: ≤5 ppm
heavy metals (as Pb): <5 ppm

application(s)

life science and biopharma
microbiology
sample preservation

foreign activity

DNase, RNase, NICKase, and protease, none detected

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol is odourless, colorless, viscous in nature, and exists as a sweet tasting liquid. It can be derived naturally as well as from petrochemical feedstock. Glycerol has a wide variety of applications, and is one of the most valuable and versatile chemical substances in nature. It can be used as an emollient, solvent, sweetening agent, in pharmaceutical formulations, cosmetics, foodstuffs and toiletries. It is very stable and can be easily stored under normal temperature; also it is non-irritating and has no adverse impact to the environment.

Application

Glycerol has been used as:
  • a component of mounting medium for immunofluorescence
  • a supplement during cell culture of Mycobacterium tuberculosis and Mycobacterium avium.
  • a fuel during the designing of enzymatic biofuel cells.
  • a liquid composite matrix with 4-HCCA and 3-amino­quinoline for analysis of neutral and acidic glycans.
  • a matrix for fast atom bombardment MS.
Glycerol may be employed as a liquid matrix for the quantification studies by MALDI (Matrix-assisted laser desorption/ionization mass spectrometry) analysis.

Biochem/physiol Actions

Glycerol is hygroscopic in nature and is soluble in water owing to its three hydrophilic alcoholic hydroxyl groups. It can form both inter- and intramolecular hydrogen bonds, making it a very flexible molecule. The physiologic effect of glycerine is due to cell-mediated immunity, increased IgG production and increased histamine release.

Features and Benefits

This product has been analyzed for the presence of nucleases and proteases.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
The product is intended for laboratory use only, and it is not suitable for drug, household, or other purposes.

greener alternative product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of glycerol/O 2 biofuel cell
Arechederra, Robert L., Becky L. Treu, and Shelley D. Minteer
Journal of Power Sources, 173 (1), 156-161 (2007)
Nur Arifah Ismail et al.
Materials science & engineering. C, Materials for biological applications, 103, 109770-109770 (2019-07-28)
In this work, the potential of titanium dioxide nanoparticles incorporated gellan gum (GG + TiO2-NPs) biofilm as wound dressing material was investigated. The GG + TiO2-NPs biofilm was prepared via evaporative casting technique and was characterized using FTIR, XRD, and SEM to study their
Comparison of the sensitivities of various derivatives of oligosaccharides in LC/MS with fast atom bombardment and electrospray ionization interfaces
Suzuki, Shigeo, Kazuaki Kakehi, and Susumu Honda
Analytical Chemistry, 68 (13) , 2073-2083 (1996)
Ionic-liquid matrices for quantitative analysis by MALDI-TOF mass spectrometry
Li, Ying L., and Michael L. Gross
Journal of the American Society For Mass Spectrometry, 15 (12), 1833-1837 (2004)
Analysis of native milk oligosaccharides directly from thin-layer chromatography plates by matrix-assisted laser desorption/ionization orthogonal-time-of-flight mass spectrometry with a glycerol matrix
Dreisewerd, Klaus, et al
Journal of the American Society For Mass Spectrometry, 17(2) , 139-150 (2006)

Protocols

Calcium phosphate transfection is a common method for the introduction of DNA into eukaryotic cells. This protocol can be optimized for use with a wide variety of cell types.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service