Skip to Content
Merck
All Photos(1)

Key Documents

T5516

Sigma-Aldrich

3,3′,5-Triiodo-L-thyronine sodium salt

γ-irradiated, powder, suitable for, suitable for cell culture

Synonym(s):

Liothyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt, T3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11I3NNaO4
CAS Number:
Molecular Weight:
672.96
Beilstein:
8179867
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.77
biological source:
synthetic (organic)
form:
powder
Assay:
≥90% (HPLC)
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

sterility

γ-irradiated

Assay

≥90% (HPLC)

form

powder

technique(s)

cell culture | mammalian: suitable

suitability

suitable for

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].N[C@@H](Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1)C([O-])=O

InChI

1S/C15H12I3NO4.Na/c16-9-6-8(1-2-13(9)20)23-14-10(17)3-7(4-11(14)18)5-12(19)15(21)22;/h1-4,6,12,20H,5,19H2,(H,21,22);/q;+1/p-1/t12-;/m0./s1

InChI key

SBXXSUDPJJJJLC-YDALLXLXSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

3,3′,5′-Triiodo-L-thyronine (T3) is an endocrine hormone produced by the thyroid gland and is regarded as an active thyroid hormone at cellular level.[1][2] Thyroxine (T4) serves as a precursor for T3 and is generated in extrathyroidal tissues.[3]

Application

3,3′,5-Triiodo-L-thyronine sodium salt has been used:
  • as a supplement in colon organ culture media[4]
  • to stimulate HuH-7 cells for analysing the hormonal influence on MBL synthesis[5]
  • to study its effect on human growth hormone (GH) versus mouse GH expression in the presence of ′′intact′′ GH genes and in a common pituitary cell context[6]

Biochem/physiol Actions

3,3′,5′-Triiodo-L-thyronine (T3) is a thyroid hormone that increases rates of proteins synthesis, stimulates the breakdown of cholesterol, and effects embryonic development. In cell culture, T3 regulates cell differentiation and protein expression. The interaction of T3 with nonhistone proteins in the chromatin initiates the thyroid hormone activity. Thyroid hormones regulate the differentiation and development of tissues and affect a number of metabolic processes.[7] The level of circulating T3 hormone is reduced in hypothyroidism.[1]

Physical form

powder-0 °C; stock-frozen in working aliquots, avoid repeated freeze/thaw, solution stable 2-8 °C, 30 days

Preparation Note

Package size based on 3,3′,5-Triiodo-L-thyronine sodium salt content; vial may also contain inert salts, such as sodium chloride

Reconstitution

To prepare 20 μg/ml stock solution: add 1 ml 1 N NaOH; gently swirl to dissolve; add 49 ml sterile medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Medical Biochemistry, 769-769 (2001)
Handbook of Behavior, Food and Nutrition, 2109-2109 (2011)
Transgenic mice expressing the human growth hormone gene provide a model system to study human growth hormone synthesis and secretion in non-tumor-derived pituitary cells: differential effects of dexamethasone and thyroid hormone
Vakili H, et al.
Molecular and cellular endocrinology, 345(1-2), 48-57 (2011)
C M Sørensen et al.
Clinical and experimental immunology, 145(1), 173-182 (2006-06-24)
Activation of the complement system via the plasma protein mannan-binding lectin (MBL) provides a first line of defence against infections. The plasma level of MBL is, in part, determined genetically, but may also be influenced by different hormones in vivo.
Irene Antolino-Lobo et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(7), 1105-1112 (2010-04-15)
Metabolism plays an important role in the toxic effects caused by 3,4-methylenedioxymethamphetamine (MDMA). Most research has focused on the involvement of CYP2D6 enzyme in MDMA bioactivation, and less is known about the contribution of other cytochrome P450 (P450) and phase

Articles

Derivation and characterization of functional human neural stem cell derived oligodendrocyte progenitor cells (OPCs) that efficiently myelinate primary neurons in culture.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service