Skip to Content
Merck
All Photos(1)

Key Documents

T2501

Sigma-Aldrich

L-Thyroxine sodium salt pentahydrate

≥98% (HPLC), powder

Synonym(s):

3-[4-(4-Hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]-L-alanine sodium salt, Sodium levothyroxine, T4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2I2OC6H2I2CH2CH(NH2)COONa · 5H2O
CAS Number:
Molecular Weight:
888.93
Beilstein:
5721845
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32
biological source:
synthetic (organic)
form:
powder

biological source

synthetic (organic)

sterility

non-sterile

Assay

≥98% (HPLC)

form

powder

mp

207-210 (dec.) (lit.)

solubility

4 M NH4OH in methanol: 50 mg/mL, clear to hazy, colorless to yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

O.O.O.O.O.[Na+].N[C@@H](Cc1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1)C([O-])=O

InChI

1S/C15H11I4NO4.Na.5H2O/c16-8-4-7(5-9(17)13(8)21)24-14-10(18)1-6(2-11(14)19)3-12(20)15(22)23;;;;;;/h1-2,4-5,12,21H,3,20H2,(H,22,23);;5*1H2/q;+1;;;;;/p-1/t12-;;;;;;/m0....../s1

InChI key

JMHCCAYJTTWMCX-QWPJCUCISA-M

Gene Information

mouse ... Cnr2(12802)
rat ... Cnr1(25248)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Thyroxine (T4) and triiodo-L-thyronine (T3) are iodine-containing hormones produced from thyroglobulin in the thyroid follicular cells. The stimulation of metabolic rate and regulation of growth and development by these hormones appear to be due to their effects on DNA transcription and thus, protein synthesis.

Preparation Note

L-Thyroxine sodium salt pentahydrate dissolves in 4 N NH4OH in methanol at 50 mg/ml to yield a clear to hazy, colorless to yellow solution.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Thyroid,Cardio-vascular system,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahdokht Jouiaei et al.
Molecular biology and evolution, 32(6), 1598-1610 (2015-03-12)
Despite Cnidaria (sea anemones, corals, jellyfish, and hydroids) being the oldest venomous animal lineage, structure-function relationships, phyletic distributions, and the molecular evolutionary regimes of toxins encoded by these intriguing animals are poorly understood. Hence, we have comprehensively elucidated the phylogenetic
Giuseppe Ianiri et al.
mBio, 6(2) (2015-04-02)
Fungal diseases represent a major burden to health care globally. As with other pathogenic microbes, there is a limited number of agents suitable for use in treating fungal diseases, and resistance to these agents can develop rapidly. Cryptococcus neoformans is
Pelin Sahlén et al.
Genome biology, 16, 156-156 (2015-08-28)
Although the locations of promoters and enhancers have been identified in several cell types, we still have limited information on their connectivity. We developed HiCap, which combines a 4-cutter restriction enzyme Hi-C with sequence capture of promoter regions. Applying the
Naoomi Tominaga et al.
Nature communications, 6, 6716-6716 (2015-04-02)
Brain metastasis is an important cause of mortality in breast cancer patients. A key event during brain metastasis is the migration of cancer cells through blood-brain barrier (BBB). However, the molecular mechanism behind the passage through this natural barrier remains
Tais A P F Doll et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(7), 1705-1713 (2015-06-09)
Successful vaccine development remains a huge challenge for infectious diseases such as malaria, HIV and influenza. As a novel way to present antigenic epitopes to the immune system, we have developed icosahedral self-assembling protein nanoparticles (SAPNs) to serve as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service