Skip to Content
Merck
All Photos(4)

Documents

H9773

Sigma-Aldrich

Hygromycin B from Streptomyces hygroscopicus

lyophilized powder, suitable for plant cell culture, BioReagent

Synonym(s):

Hygromycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37N3O13
CAS Number:
Molecular Weight:
527.52
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.72

product name

Hygromycin B from Streptomyces hygroscopicus, suitable for plant cell culture, BioReagent, ≥60% (HPLC), lyophilized powder

biological source

Streptomyces hygroscopicus

Quality Level

product line

BioReagent

form

lyophilized powder

purified by

ion-exchange chromatography

concentration

≥60% (HPLC)

technique(s)

cell culture | plant: suitable

color

faintly brown to brown
white to beige

 

(1) 7.1, (2) 8.8

solubility

H2O: soluble 50 mg/mL
ethanol: soluble
methanol: soluble

antibiotic activity spectrum

fungi

application(s)

agriculture

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CN[C@H]1C[C@@H](N)[C@H](O)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@@H]3O[C@]4(O[C@H]([C@H](N)CO)[C@H](O)[C@H](O)[C@H]4O)O[C@H]23)[C@@H]1O

InChI

1S/C20H37N3O13/c1-23-7-2-5(21)9(26)15(10(7)27)33-19-17-16(11(28)8(4-25)32-19)35-20(36-17)18(31)13(30)12(29)14(34-20)6(22)3-24/h5-19,23-31H,2-4,21-22H2,1H3/t5-,6-,7+,8-,9+,10-,11+,12-,13+,14-,15-,16+,17+,18-,19+,20+/m1/s1

InChI key

GRRNUXAQVGOGFE-XKIAHZFYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside
Hygromycin B is an antibiotic produced by the bacterium Streptomyces hygroscopicus. It is an aminoglycoside that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis.

Application

Hygromycin B is an aminoglycoside antibiotic isolated from Streptomyces hygroscopicus. It has been used to study protein synthesis at the level of the 70S ribosome translocation and mRNA template misreading, as an antiviral agent by selectively penetrating cells rendered permeable by virus infection and inhibiting translation, and as a selection agent for hygromycin resistance gene transformed cells. It is recommended for use as a selection agent at 100-800 μg/mL, specifically at 100 μg/mL for prokaryotes, 200 μg/mL for lower eukaryotes and 150-400 μg/mL for higher eukaryotes.

Hygromycin B from Streptomyces hygroscopicus is used for gene cloning and ectopic expression. It is also used in MS medium for the in vitro screening of transgenic plants.

Biochem/physiol Actions

Hygromycin B is used in mutagenesis or for selection of gene expression and complementation constructs in C. jejuni. It is also adopted as a marker in other approaches for manipulating, introducing or deleting DNA in C. elegans. It can be inactivated by Aminoglycoside 4-phosphotransferase-Ia (APH(4)-Ia)/Hygromycin B phosphotransferase (Hph) through phosphorylation. The amino function of this antibiotic may be involved in a general acid-base catalysis performed by the ribozyme, acting as proton acceptors/donors.
Mode of Action: The product acts by inhibiting protein synthesis by inducing the misreading of the m-RNA template in the prokaryote, with the potency to inhibit translation.

Antimicrobial Spectrum: Hygromycin B acts against bacteria, fungi and higher eukaryotic cells.

Caution

Hygromycin B products should be stored as supplied at 2-8°C, and the dry solid is table for at least 5 years if stored at 2-8°C. It is stable at 37°C for 30 days. (For solutions) This solution is stable as supplied for two years if stored at 2-8°C.

Preparation Note

This product is purified by ion exchange chromatography. Hygromycin B is soluble in water at concentrations >50 mg/mL, and also soluble in methanol or ethanol. Solutions should be sterilized by filtration, not by autoclaving. Additionally, Hygromycin B solutions have been reported to lose activity on freezing, and since they are stable refrigerated, freezing should be avoided.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yajun Xi et al.
Methods in molecular biology (Clifton, N.J.), 581, 53-59 (2009-09-22)
Switchgrass (Panicum virgatum L.) is a highly productive warm-season C4 species that is being developed into a dedicated biofuel crop. This chapter describes a protocol that allows the generation of transgenic switchgrass plants by Agrobacterium tumefaciens-mediated transformation. Embryogenic calluses induced
M Dutt et al.
Plant cell reports, 29(11), 1251-1260 (2010-08-17)
A method for the genetic transformation of several citrus cultivars is described, including cultivars observed to be recalcitrant to conventional epicotyl-mediated transformation. Embryogenic cell suspension cultures, established from unfertilized ovules were used as target tissues for Agrobacterium-mediated transformation. Several modifications
Tsuyoshi Nakagawa et al.
Journal of bioscience and bioengineering, 104(1), 34-41 (2007-08-19)
We developed a new series of binary vectors useful for Gateway cloning to facilitate transgenic experiments in plant biotechnology. The new system, Gateway Binary Vectors (pGWBs) realized efficient cloning, constitutive expression using the cauliflower mosaic virus (CaMV) 35S promoter and
Inja Radman et al.
PloS one, 8(10), e76019-e76019 (2013-10-17)
We report a simple, cost-effective, scalable and efficient method for creating transgenic Caenorhabditis elegans that requires minimal hands-on time. The method combines biolistic bombardment with selection for transgenics that bear a hygromycin B resistance gene on agar plates supplemented with
Andrew Cameron et al.
PloS one, 9(4), e95084-e95084 (2014-04-23)
Campylobacter jejuni genetic manipulation is restricted by the limited number of antibiotic resistance cassettes available for use in this diarrheal pathogen. In this study, two antibiotic resistance cassettes were developed, encoding for hygromycin B and apramycin resistance, for use in

Articles

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

Prevent fungal, yeast, and mold contamination in cell cultures. Discover the best antifungal agent for your cultures with the extensive Sigma® antifungal collection.

See All

Protocols

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service