Skip to Content
Merck
All Photos(1)

Documents

45684

Supelco

Thiabendazole

PESTANAL®, analytical standard

Synonym(s):

2-(4-Thiazolyl)benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7N3S
CAS Number:
Molecular Weight:
201.25
Beilstein:
611403
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E233
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

fungi

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc2[nH]c(nc2c1)-c3cscn3

InChI

1S/C10H7N3S/c1-2-4-8-7(3-1)12-10(13-8)9-5-14-6-11-9/h1-6H,(H,12,13)

InChI key

WJCNZQLZVWNLKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stuart Dashper et al.
Antimicrobial agents and chemotherapy, 54(3), 1311-1314 (2009-12-30)
Porphyromonas gingivalis is a major pathogen of chronic periodontitis and exists in a biofilm on the surface of the tooth root. Oxantel, a cholinergic anthelmintic and fumarate reductase inhibitor, significantly inhibited biofilm formation by P. gingivalis and disrupted established biofilms
Matthieu Groh et al.
Clinical and experimental rheumatology, 30(1 Suppl 70), S107-S109 (2012-07-26)
Polyarteritis nodosa (PAN), a systemic necrotising vasculitis that affects medium- and small-sized arteries, has visceral involvement in 40-60% of the patients. According to the Five-Factor Score (FFS), it is associated with poor outcome. We describe a patient who underwent orthotopic
Andrea Peano et al.
Veterinary dermatology, 23(2), 131-135 (2012-02-09)
In this study, we evaluated the antifungal susceptibility of Malassezia pachydermatis to clotrimazole (CTZ), miconazole (MCZ), and thiabendazole (TBD), azole derivatives employed in aural formulations labeled for treatment of canine otitis. The procedure for in vitro testing was based on
P Fonderie et al.
Parasitology, 139(10), 1301-1308 (2012-06-22)
Studies on anthelmintic resistance in equine parasites do not include facultative parasites. Halicephalobus gingivalis is a free-living bacterivorous nematode and a known facultative parasite of horses with a strong indication of some form of tolerance to common anthelmintic drugs. This
Hye Ji Cha et al.
PLoS biology, 10(8), e1001379-e1001379 (2012-08-29)
Studies in diverse organisms have revealed a surprising depth to the evolutionary conservation of genetic modules. For example, a systematic analysis of such conserved modules has recently shown that genes in yeast that maintain cell walls have been repurposed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service