Skip to Content
Merck
All Photos(1)

Key Documents

T2379

Sigma-Aldrich

Tubocurarine hydrochloride pentahydrate

≥97%

Synonym(s):

(+)-Tubocurarine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H41ClN2O6 · HCl · 5H2O
CAS Number:
Molecular Weight:
771.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97%

solubility

H2O: soluble 50 mg/ml, with heating as required, clear to slightly hazy, colorless to yellow
ethanol: soluble
methanol: soluble

storage temp.

2-8°C

SMILES string

O.O.O.O.O.Cl.[Cl-].COc1cc2CCN(C)[C@H]3Cc4ccc(Oc5c(O)c(OC)cc6CC[N+](C)(C)[C@H](Cc7ccc(O)c(Oc1cc23)c7)c56)cc4

InChI

1S/C37H40N2O6.2ClH.5H2O/c1-38-14-12-24-19-32(42-4)33-21-27(24)28(38)16-22-6-9-26(10-7-22)44-37-35-25(20-34(43-5)36(37)41)13-15-39(2,3)29(35)17-23-8-11-30(40)31(18-23)45-33;;;;;;;/h6-11,18-21,28-29H,12-17H2,1-5H3,(H-,40,41);2*1H;5*1H2/t28-,29+;;;;;;;/m0......./s1

InChI key

WMIZITXEJNQAQK-GGDSLZADSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tubocurarine hydrochloride pentahydrate has been used as a acetylcholinesterase inhibitor (Ach) inhibitor in myotubes, muscle and as neuromuscular junction blocker in zebra fish embryos.

Biochem/physiol Actions

Tubocurarine is a muscle relaxant and a nicotinic acetylcholine receptor antagonist. It can induce neuromuscular blocking.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Tubocurarine hydrochloride pentahydrate is soluble in water at 50 mg/ml (with heat as required) and yields a clear to slightly hazy, colorless to yellow solution. It is also soluble in methanol and ethanol.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerta Vrbova et al.
The European journal of neuroscience, 30(3), 366-375 (2009-08-07)
There are receptors on denervated Schwann cells that may respond to the neurotransmitters that are released from growth cones of regenerating motor axons. In order to ascertain whether the interaction of the transmitters and their receptors plays a role during
Proteasome inhibition with bortezomib depletes plasma cells and autoantibodies in experimental autoimmune myasthenia gravis
Muto A, et al.
Proceedings of the National Academy of Sciences of the USA, 108(13), 5425-5425 (2011)
A Ashkenazi et al.
Journal of the neurological sciences, 164(2), 124-128 (1999-07-13)
The pharmacology of auditory brain-stem evoked potentials (ABEP) pathways is poorly understood. There are anecdotal reports on the involvement of various neurotransmitters but they were not investigated systematically. The aim of this study was to investigate the effects on ABEP
Matthew Lovett-Barron et al.
Nature neuroscience, 23(8), 959-967 (2020-06-24)
The hypothalamus is composed of many neuropeptidergic cell populations and directs multiple survival behaviors, including defensive responses to threats. However, the relationship between the peptidergic identity of neurons and their roles in behavior remains unclear. Here, we address this issue
Proteasome inhibition with bortezomib depletes plasma cells and autoantibodies in experimental autoimmune myasthenia gravis
Gomez A, et al.
Journal of immunology (Baltimore, Md. : 1950), 186(4), 2503-2513 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service