Skip to Content
Merck
All Photos(1)

Key Documents

A2661

Sigma-Aldrich

Acetylcholine chloride

suitable for cell culture

Synonym(s):

ACh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N+CH2CH2OCOCH3Cl-
CAS Number:
Molecular Weight:
181.66
Beilstein:
3571875
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32190102
PubChem Substance ID:
NACRES:
NA.75
biological source:
synthetic
form:
powder
Assay:
~99%
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

~99%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

146-150 °C (lit.)

solubility

water: 100 mg/mL, clear, colorless

shipped in

ambient

storage temp.

room temp

SMILES string

[Cl-].CC(=O)OCC[N+](C)(C)C

InChI

1S/C7H16NO2.ClH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1

InChI key

JUGOREOARAHOCO-UHFFFAOYSA-M

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

Application

Acetylcholine chloride has been used in the preparation of acetylcholine, that is used as a common vehicle for iontophoresis of acetylcholine (ACh).[1] It has also been used to treat aNS-1 derived neurons for cAMP (cyclic adenosine monophosphate) assay and also used to check their ability to influence G protein-coupled receptors (GPCRs).[2]

Biochem/physiol Actions

Acetylcholine chloride, injected at 20 mg/kg body weight, reduces mortality and plasma proinflammatory cytokines in mice with experimentally-induced sepsis . The cholinergic anti-inflammatory mechanism is probably mediated by interaction of acetylcholine with α7n cholinoreceptor on monocytes, macrophages, and neutrophils, which decreases the levels of proinflammatory cytokines such as TNF-α, IL-1β, and IL-6.
Endogenous neurotransmitter at cholinergic synapses; amplifies action potential of the sarcolemma thereby inducing muscle contractions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel gel based vehicle for the delivery of acetylcholine in quantitative sudomotor axon reflex testing
Sletten D M, et al.
Autonomic Neuroscience : Basic & Clinical, 150(1-2), 127-130 (2009)
Adaptation of NS cells growth and differentiation to high-throughput screening-compatible plates
Garavaglia A, et al.
BMC Neuroscience, 11(1), 7-7 (2010)
Yue-Chen Chang et al.
Kidney & blood pressure research, 43(5), 1607-1622 (2018-10-23)
This experimental study aims to observe whether the protective effect of propofol against renal ischemia-reperfusion injury (IRI) in the rat interlobar artery occurs through altered expression of the gap junction protein connexin 43 (Cx43). This study randomly divided male Sprague
Wei Shi et al.
The Journal of biological chemistry, 295(22), 7653-7668 (2020-04-24)
The erythropoietin-producing human hepatocellular receptor EPH receptor B6 (EPHB6) is a receptor tyrosine kinase that has been shown previously to control catecholamine synthesis in the adrenal gland chromaffin cells (AGCCs) in a testosterone-dependent fashion. EPHB6 also has a role in
Fabian Hertel et al.
ACS chemical biology, 15(1), 33-38 (2019-12-20)
Phosphoinositides constitute a critical family of lipids that regulate numerous cellular processes. Phosphatidylinositol 4,5-bisphosphate (PIP2) is arguably the most important plasma membrane phosphoinositide and is involved in regulating diverse processes. It is also the precursor of phosphatidylinositol 3,4,5-trisphosphate (PIP3), which

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service