Skip to Content
Merck
All Photos(1)

Key Documents

SML1743

Sigma-Aldrich

ETaKG

≥95% (HPLC)

Synonym(s):

2-Oxo-pentanedioic acid 5-ethyl ester 1-(3-trifluoromethyl-benzyl) ester, 5-Ethyl 1-(3-(trifluoromethyl)benzyl) 2-oxopentanedioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15F3O5
CAS Number:
Molecular Weight:
332.27
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

oil

color

colorless to light yellow

storage temp.

2-8°C

SMILES string

O=C(C(CCC(OCC)=O)=O)OCC1=CC=CC(C(F)(F)F)=C1

InChI

1S/C15H15F3O5/c1-2-22-13(20)7-6-12(19)14(21)23-9-10-4-3-5-11(8-10)15(16,17)18/h3-5,8H,2,6-7,9H2,1H3

InChI key

OKDCLZOKKGLMIX-UHFFFAOYSA-N

Biochem/physiol Actions

A cell-permeable, bioavailable & aqueous stable α-ketoglutarate prodrug that selectively induces PHD-dependent cell death in hypoxic cells
ETaKG is a cell-permeable, aqueous stable, bioavailable, non-toxic α-ketoglutarate/2-oxoglutarate (α-KG/2-OG) diester derivative that readily undergoes hydrolysis by cytosolic esterases to α-KG. The monoester analog, TaKG is also membrane permeant. ETaKG elevates intracellular [α-KG] by ~15-fold in a prologned fashion, destabilizes HIF-1α/2α and reactivates prolyl hydroxylases (PHDs) in hypoxic HCT116, A431 and RPE cells at 2 mM. Further, shown to selectively trigger PHD-dependent cell death in hypoxic cells and decreases cellular [ATP] and glucose uptake. ETaKG has shown to reduce glucose metabolism, elevate [α-KG] levels and suppress A375 xenograft tumor growth (750 mg/kg of mice, p.o., q.d., 14 days). Also, induces mTORC1 lysosomal translocation and stimulates mTORC1 activity in the absence of glutaminolysis in U2OS cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanumantha Rao Madala et al.
Cancer research, 80(17), 3492-3506 (2020-07-12)
Under conditions of inherent or induced mitochondrial dysfunction, cancer cells manifest overlapping metabolic phenotypes, suggesting that they may be targeted via a common approach. Here, we use multiple oxidative phosphorylation (OXPHOS)-competent and incompetent cancer cell pairs to demonstrate that treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service