Skip to Content
Merck
All Photos(1)

Documents

349631

Sigma-Aldrich

Dimethyl 2-oxoglutarate

96%

Synonym(s):

Dimethyl α-ketoglutarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2CCH2CH2COCO2CH3
CAS Number:
Molecular Weight:
174.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

90-95 °C/0.4 mmHg (lit.)

density

1.203 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCC(=O)C(=O)OC

InChI

1S/C7H10O5/c1-11-6(9)4-3-5(8)7(10)12-2/h3-4H2,1-2H3

InChI key

TXIXSLPEABAEHP-UHFFFAOYSA-N

General description

Dimethyl 2-oxoglutarate is a key intermediate formed during the Krebs cycle and an important nitrogen transporter in the biological metabolic pathways. The electrochemical behavior of dimethyl-2-oxoglutarate has been investigated by cyclic voltammetry, square wave voltammetry and differential pulse voltammetry using a glassy carbon electrode.

Application

Dimethyl 2-oxoglutarate can undergo cylocondensation with dinucleophiles, such as 1,2-phenylenediamine, 2-aminophenol and 2-aminobenzenethiol to form novel heterocycles.
Dimethyl 2-oxoglutarate may be used to synthesize the conformationally constrained PNA (peptide nucleic acid) -monomer capable of binding thymine in a triplex motif. It may be used in the synthesis of 4-aryl kainic acid analogs, via highly stereoselective Michael addition reaction with nitrostyrene.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Afzal Shah et al.
Bioelectrochemistry (Amsterdam, Netherlands), 77(2), 145-150 (2009-09-22)
The electrochemical behaviour of dimethyl-2-oxoglutarate (MOG), a key intermediate in the Krebs cycle and an important nitrogen transporter in the metabolic pathways in biological processes, was investigated by cyclic voltammetry, square wave voltammetry and differential pulse voltammetry using a glassy
Organic syntheses based on 2?oxoglutaric acid. V. Syntheses of novel 2H?1, 4?benzothiazines and a 2, 5?dihydro?1, 5?benzothiazepine.
Blitzke T, et al.
Journal of Heterocyclic Chemistry, 34(2), 453-455 (1997)
Daniela Gaglio et al.
Molecular systems biology, 7, 523-523 (2011-08-19)
Oncogenes such as K-ras mediate cellular and metabolic transformation during tumorigenesis. To analyze K-Ras-dependent metabolic alterations, we employed ¹³C metabolic flux analysis (MFA), non-targeted tracer fate detection (NTFD) of ¹⁵N-labeled glutamine, and transcriptomic profiling in mouse fibroblast and human carcinoma
Yuan Cao et al.
Theranostics, 7(12), 3021-3033 (2017-08-26)
Increased glutamine metabolism is a hallmark of cancer. Mitochondrial glutamic pyruvate transaminase (GPT2) catalyzes the reversible transamination between alanine and α-ketoglutarate (α-KG), also known as 2-oxoglutarate, to generate pyruvate and glutamate during cellular glutamine catabolism. However, the precise role of
An efficient synthesis of 4-aryl kainic acid analogs.
Maeda H, et al.
Tetrahedron, 55(4), 943-954 (1999)

Articles

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Sigma article discusses tumor cell metabolic pathways, focusing on aerobic glycolysis and mitochondrial activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service