Skip to Content
Merck
All Photos(1)

Key Documents

567375

Sigma-Aldrich

Smac-N7 Peptide, Cell-Permeable

The Smac-N7 peptide is modified to be cell permeant by linking the lysine carboxyl terminal to the arginine of Antennapedia homeodomain 16-mer peptide via a proline linker.

Synonym(s):

Smac-N7 Peptide, Cell-Permeable, AVPIAQKPRQIKIWFQNRRMKWKK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C142H232N44O29S
Molecular Weight:
3051.70
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

The Smac-N7 peptide (Cat. No. 567370) is modified to be cell permeant by linking the lysine carboxyl terminal to the arginine of Antennapedia homeodomain 16-mer peptide via a proline linker.
The Smac-N7 peptide (Cat. No. 567370) where the lysine carboxy terminal is linked to the arginine of Antennapedia homeodomain 16-mer peptide (underlined) via a proline linker to make it cell-permeable. Smac-N7 peptide contains the N-terminal seven residues of Smac and promotes the activation of procaspase-3 (~10 µM). Also, promotes the catalytic activity of caspase-9 in the presence of XIAP.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Promotes procaspase-3 activation
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Ala-Val-Pro-Ile-Ala-Gln-Lys-Pro-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Srinivasula, S.M., et al. 2001. Nature410, 112.
Chai, J., et al. 2000. Nature406, 855.
Liu, Z., et al. 2000. Nature408, 1004.
Srinivasula, S.M., et al. 2000. J. Biol. Chem.275, 36152.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service