Skip to Content
Merck
All Photos(1)

Key Documents

SML0652

Sigma-Aldrich

IOX2

≥98% (HPLC)

Synonym(s):

JICL38, N-[[1,2-Dihydro-4-hydroxy-2-oxo-1-(phenylmethyl)-3-quinolinyl]carbonyl]-glycine, N-[[4-Hydroxy-2-oxo-1-(phenylmethyl)-1,2-dihydro-3-quinolinyl]carbonyl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16N2O5
CAS Number:
Molecular Weight:
352.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC(C1=C(N2CC3=CC=CC=C3)C=CC=C1)=C(C(NCC(O)=O)=O)C2=O

InChI

1S/C19H16N2O5/c22-15(23)10-20-18(25)16-17(24)13-8-4-5-9-14(13)21(19(16)26)11-12-6-2-1-3-7-12/h1-9,24H,10-11H2,(H,20,25)(H,22,23)

InChI key

CAOSCCRYLYQBES-UHFFFAOYSA-N

Application

IOX2 has been used as PHD inhibitor to induce pseudo-hypoxia.

Biochem/physiol Actions

IOX2 is selective and potent inhibitor of prolyl hydroxylases (PHD). Stabilization of HIF-1R through inhibition of PHD has been examined as a potential treatment for ischemic diseases including anemia, myocardial infarction, and stroke. IOX2 is a selective inhibitor of the hypoxia inducible factor (HIF) prolyl-hydroxylases (PHD) that exhibits up-regulation of HIF1a in zebrafish. For full characterization details, please visit the IOX2 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

IOX2 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resveratrol integrates metabolic and growth effects in PC3 prostate cancer cells-involvement of prolyl hydroxylase and hypoxia inducible factor-1
Fonseca J, et al.
Oncology Letters (2018)
Pieter-Jan Stiers et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 34(2), 333-348 (2018-11-20)
Tissue engineering holds great promise for bone regenerative medicine, but clinical translation remains challenging. An important factor is the low cell survival after implantation, primarily caused by the lack of functional vasculature at the bone defect. Interestingly, bone development and
Hu Peng et al.
American journal of physiology. Renal physiology, 318(2), F468-F474 (2019-12-17)
Acute pyelonephritis is frequently associated with metabolic acidosis. We previously reported that metabolic acidosis stimulates expression of hypoxia-inducible factor (HIF)-1α-induced target genes such as stromal derived factor-1 and cathelicidin, an antimicrobial peptide. Since the collecting duct (CD) plays a pivotal
Deirdre Scully et al.
Development (Cambridge, England), 143(10), 1742-1752 (2016-05-18)
Hypoxia is encountered in either pathological or physiological conditions, the latter of which is seen in amniote embryos prior to the commencement of a functional blood circulation. During the hypoxic stage, a large number of neural crest cells arise from
Lin Zhao et al.
Molecular medicine reports, 22(1), 436-444 (2020-05-08)
Posterior capsular opacification (PCO) is the main complication following cataract surgery. The proliferation of the residual lens epithelial cells (LECs) serves an important role in PCO formation. The authors' previous study revealed that vitamin C inhibited the proliferation of human

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service