Skip to Content
Merck
All Photos(1)

Key Documents

SML1107

Sigma-Aldrich

SGC0946

≥98% (HPLC)

Synonym(s):

1-(3-((((2R,3S,4R,5R)-5-(4-Amino-5-bromo-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl)(isopropyl)amino)propyl)-3-(4-(tert-butyl)phenyl)urea, 5-Bromo-7-[5-deoxy-5-[[3-[[[[4-(1,1-dimethylethyl)phenyl]amino]carbonyl]amino]propyl](1-methylethyl)amino]-beta-D-7H-Pyrrolo[2,3-d]pyrimidin-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H40BrN7O4
CAS Number:
Molecular Weight:
618.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

BrC1=CN([C@@H]2O[C@H](CN(CCCNC(NC3=CC=C(C(C)(C)C)C=C3)=O)C(C)C)[C@@H](O)[C@H]2O)C4=C1C(N)=NC=N4

InChI

1S/C28H40BrN7O4/c1-16(2)35(12-6-11-31-27(39)34-18-9-7-17(8-10-18)28(3,4)5)14-20-22(37)23(38)26(40-20)36-13-19(29)21-24(30)32-15-33-25(21)36/h7-10,13,15-16,20,22-23,26,37-38H,6,11-12,14H2,1-5H3,(H2,30,32,33)(H2,31,34,39)/t20-,22-,23-,26-/m1/s1

InChI key

IQCKJUKAQJINMK-HUBRGWSESA-N

Biochem/physiol Actions

SGC0946 is a potent and selective inhibitor of Histone H3-lysine79 (H3K79) methyltransferase DOT1L with an IC50 of 0.3 nM in a radioactive enzyme assay, and over 100-fold selectivity for DOT1L over other histone methyltransferases. In cell studies, SGC0946 reduces H3K79 dimethylation with an IC50 of 2.6 nM in A431 cells and 8.8 nM in MCF10A cells. SGC0946 was found to selectively kills cells transformed with the MLL-AF9 fusion oncogene in an in vitro model of leukemia. For full characterization details, please visit the SGC0946 probe summary on the Structural Genomics Consortium (SGC) website.

SGC0649 is the negative control for the active probe, SGC0946. To request a sample of the negative control from the SGC, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
SGC0946 is a potent and selective inhibitor of Histone H3-lysine79 (H3K79) methyltransferase DOT1L.

Features and Benefits

SGC0946 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

SGC0946 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the SGC0946 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Zhang et al.
eLife, 9 (2020-10-02)
Aberrant HOXA9 expression is a hallmark of most aggressive acute leukemias, notably those with KMT2A (MLL) gene rearrangements. HOXA9 overexpression not only predicts poor diagnosis and outcome but also plays a critical role in leukemia transformation and maintenance. However, our
Julia Gobl et al.
Vaccines, 8(2) (2020-04-05)
Epigenetic mechanisms have not been characterized in ticks despite their importance as vectors of human and animal diseases worldwide. Our investigation identifies and functionally characterizes the orthologue of S-adenosylmethionine (SAM) binding methyltransferase enzyme, disruptor of telomeric silencing 1-like (DOT1L) in
Kazuya Ishiguro et al.
Haematologica, 104(1), 155-165 (2018-09-02)
Epigenetic alterations play an important role in the pathogenesis in multiple myeloma, but their biological and clinical relevance is not fully understood. Here, we show that DOT1L, which catalyzes methylation of histone H3 lysine 79, is required for myeloma cell
Giovanni Nassa et al.
Science advances, 5(2), eaav5590-eaav5590 (2019-02-19)
Breast cancer (BC) resistance to endocrine therapy results from constitutively active or aberrant estrogen receptor α (ERα) signaling, and ways to block ERα pathway in these tumors are sought after. We identified the H3K79 methyltransferase DOT1L as a novel cofactor
Christiaan J Stavast et al.
Leukemia (2021-11-07)
MIR139 is a tumor suppressor and is commonly silenced in acute myeloid leukemia (AML). However, the tumor-suppressing activities of miR-139 and molecular mechanisms of MIR139-silencing remain largely unknown. Here, we studied the poorly prognostic MLL-AF9 fusion protein-expressing AML. We show

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service