Skip to Content
Merck
All Photos(1)

Key Documents

D9533

Sigma-Aldrich

Deferoxamine mesylate salt

≥92.5% (TLC), powder, iron-chelating agent

Synonym(s):

DFX mesylate, DFOM, Deferoxamine methanesulfonate salt, Desferrioxamine mesylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H48N6O8 · CH4O3S
CAS Number:
Molecular Weight:
656.79
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

Product Name

Deferoxamine mesylate salt, powder, ≥92.5% (TLC)

Quality Level

Assay

≥92.5% (TLC)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

storage temp.

−20°C

SMILES string

CS(O)(=O)=O.CC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCN

InChI

1S/C25H48N6O8.CH4O3S/c1-21(32)29(37)18-9-3-6-16-27-22(33)12-14-25(36)31(39)20-10-4-7-17-28-23(34)11-13-24(35)30(38)19-8-2-5-15-26;1-5(2,3)4/h37-39H,2-20,26H2,1H3,(H,27,33)(H,28,34);1H3,(H,2,3,4)

InChI key

IDDIJAWJANBQLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Deferoxamine mesylate salt has been used



  • for cell pretreatment to assess its influenceon cell viability and its interactions with other compounds ()
  • to study the effect on Lipocalin 2(Lcn2) production triggered by amyloid-β(Aβ). ()
  • used to induce iron starvation in bacterial cultures.()

Biochem/physiol Actions

An iron chelator used often in the studies of cell proliferation and apoptosis. Also reported to induce p53.
An iron chelator used often in the studies of cell proliferation and apoptosis. Has been shown to have anti-proliferative effects on vascular smooth muscle cells in vitro and in vivo and to arrest cells in the G1 phase. Also reported to induce p53. Induces apoptosis in HL-60 cells by chelating iron. After 48 hrs treatment with 1μM deferoxamine, DNA fragmentation was apparent. Cells treated with 0.1 μM deferoxamine for as little as 24 hours were committed to apoptosis; by 48 hrs nuclear collapse was observed. In some studies it has been shown to have antioxidant properties and to protect cells against H2O2-induced damage.
Deferoxamine is used as a hypoxia-mimetic agent to stabilize Hypoxia Inducible Factor 1 (HIF-1). Deferoxamine stabilizes HIF-1 through the inhibition of Prolyl Hydroxylases (PHDs) which target HIF-1 through degradation. The mechanism of deferoxamine inhibition is likely through the chelation of Fe2+ bound to the active site of PHD which is required for enzymatic activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Porreca et al.
Arteriosclerosis and thrombosis : a journal of vascular biology, 14(2), 299-304 (1994-02-01)
Vascular smooth muscle cell (VSMC) growth is a primary component of accelerated and spontaneous atherosclerosis. Previous studies have shown that iron may be involved in the control of enzymatic activities that modulate DNA synthesis in human cells. In this study
Biodegradable hypoxia biomimicry microspheres for bone tissue regeneration.
Lina Hadidi et al.
Journal of biomaterials applications, 34(7), 1028-1037 (2019-10-28)
S Bereswill et al.
Journal of bacteriology, 182(21), 5948-5953 (2000-10-13)
Homologs of the ferric uptake regulator Fur and the iron storage protein ferritin play a central role in maintaining iron homeostasis in bacteria. The gastric pathogen Helicobacter pylori contains an iron-induced prokaryotic ferritin (Pfr) which has been shown to be
W G An et al.
Nature, 392(6674), 405-408 (1998-04-16)
Although hypoxia (lack of oxygen in body tissues) is perhaps the most physiological inducer of the wild-type p53 gene, the mechanism of this induction is unknown. Cells may detect low oxygen levels through a haem-containing sensor protein. The hypoxic state
Yuanyuan Kong et al.
Cell death & disease, 10(9), 624-624 (2019-08-20)
Recent findings demonstrate that aberrant downregulation of the iron-exporter protein, ferroportin (FPN1), is associated with poor prognosis and osteoclast differentiation in multiple myeloma (MM). Here, we show that FPN1 was downregulated in MM and that clustered regularly interspaced short palindromic

Articles

Viable but nonculturable (VBNC) bacteria describes cells that cannot be normally cultured however, “resuscitation” of these cells can be mediated by a physical and different kind of chemical stimuli.

Viable but nonculturable (VBNC) bacteria describes cells that cannot be normally cultured however, “resuscitation” of these cells can be mediated by a physical and different kind of chemical stimuli.

Viable but nonculturable (VBNC) bacteria describes cells that cannot be normally cultured however, “resuscitation” of these cells can be mediated by a physical and different kind of chemical stimuli.

Viable but nonculturable (VBNC) bacteria describes cells that cannot be normally cultured however, “resuscitation” of these cells can be mediated by a physical and different kind of chemical stimuli.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service