Skip to Content
Merck
All Photos(4)

Key Documents

HPA018111

Sigma-Aldrich

Anti-ELN antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Elastin precursor, Anti-Tropoelastin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VKPGKVPGVGLPGVYPGGVLPGARFPGVGVLPGVPTGAGVKPKAPGVGGAFAGIPGVGPFGGPQPGVPLGYPIKAPKLPGGYGLPYTTGKLPYGYGPGGVAGAAGKAGYPTGTGVGPQAAAAAAAKAAAKFGAGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELN(2006)

General description

Elastin (ELN) is a non-soluble protein. The highly durable protein is a major component of the extracellular matrix and is an intrinsic indicator of pathologic states.

Immunogen

Elastin precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ELN antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Elastin (ELN) is one protein which remains functional throughout a lifetime. It offers the critical recoil property for all tissues that deal with constant strain. Structural properties and mechanical strength of elastic tissues in the lungs, skin and vasculature are contributed by ELN. During wound healing it is deficient and once scar tissue starts forming, ELN exists as a disorganized network of fibers. Processes like cell migration and matrix synthesis are mediated by it. Mutations in the gene encoding ELN have been associated with cutis laxa and supravalvular aortic stenosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73202

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark L Siefring et al.
Pediatric dermatology, 31(3), 347-349 (2014-04-25)
We report a 3-year-old girl from Vietnam with severe congenital cutis laxa; no cardiovascular, pulmonary, neurologic, or visceral involvement; and no family history of cutis laxa. Mutational analysis of the elastin gene identified heterozygosity for a previously unreported de novo
Z Urbán et al.
Human genetics, 106(6), 577-588 (2000-08-15)
We have used single-strand conformation and heteroduplex analyses of genomic amplimers to identify point mutations within the elastin gene (ELN) in patients with non-syndromic supravalvular aortic stenosis (SVAS) from a total of eight unrelated families. Six novel point mutations were
Miriam Votteler et al.
Development (Cambridge, England), 140(11), 2345-2353 (2013-05-03)
Semilunar valve leaflets have a well-described trilaminar histoarchitecture, with a sophisticated elastic fiber network. It was previously proposed that elastin-containing fibers play a subordinate role in early human cardiac valve development; however, this assumption was based on data obtained from
Giselle C Yeo et al.
The Journal of biological chemistry, 289(50), 34815-34826 (2014-10-25)
Tropoelastin is an extracellular matrix protein that assembles into elastic fibers that provide elasticity and strength to vertebrate tissues. Although the contributions of specific tropoelastin regions during each stage of elastogenesis are still not fully understood, studies predominantly recognize the
Consuelo Pérez-Rico et al.
Investigative ophthalmology & visual science, 55(10), 6309-6318 (2014-09-04)
We evaluated the expression of several extracellular matrix constituents implicated in the synthesis and reticulation of elastin in human pterygium, according to age and sex of the patients. Pterygia and normal conjunctiva samples were divided into groups according to age

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service