Skip to Content
Merck
All Photos(2)

Key Documents

RAB0464

Sigma-Aldrich

Mouse Thrombopoietin ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

TPO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 30 pg/mL
standard curve range: 27.4-20000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Thpo(21832)

General description

The Mouse TPO ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse TPO in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse Thrombopoietin

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irene Ciancarelli et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 19(6), 452-456 (2014-07-02)
Peripheral oxidative biomarkers could be useful for monitoring clinical features of Huntington's disease (HD). Cu/Zn-superoxide dismutase (Cu/Zn-SOD), neuron-specific enolase (NSE) and 8-hydroxy-2'-deoxyguanosine (8-oxoGua) serum levels were analysed in 18 HD patients and 10 controls. Clinical measures were recorded from each
Y-L Zhang et al.
Letters in applied microbiology, 59(6), 580-586 (2014-08-29)
Clostridium sporogenes ATCC 3584 is an obligate anaerobe that has been reported to possess excellent tumour-targeting capacity. Here, we use Cl. sporogenes as a vector to deliver IL-12, a potent antitumour cytokine that bears numerous antitumour properties but that has limited
Nike Müller et al.
PloS one, 9(4), e94282-e94282 (2014-04-15)
Inflammation and metabolism have been shown to be evolutionary linked and increasing evidence exists that pro-inflammatory factors are involved in the pathogenesis of obesity and type 2 diabetes. Until now, most data suggest that within adipose tissue these factors are
Sangbin Park et al.
PLoS genetics, 10(8), e1004555-e1004555 (2014-08-08)
Insulin is a major regulator of metabolism in metazoans, including the fruit fly Drosophila melanogaster. Genome-wide association studies (GWAS) suggest a genetic basis for reductions of both insulin sensitivity and insulin secretion, phenotypes commonly observed in humans with type 2
Helena Tizón-Marcos et al.
The American journal of cardiology, 114(7), 1094-1099 (2014-08-19)
Increased myocardial trabeculations define noncompaction cardiomyopathy (NCC). Imaging advancements have led to increasingly common identification of prominent trabeculations with unknown implications. We quantified and determined the impact of trabeculations' burden on cardiac function and stretch in a population of healthy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service