Skip to Content
Merck
All Photos(2)

Key Documents

R0500

Sigma-Aldrich

Reichstein′s substance S

≥98% purity, powder

Synonym(s):

11-Deoxy-17-hydroxycorticosterone, 11-Deoxycortisol, 17α,21-Dihydroxy-4-pregnene-3,20-dione, 17α-Hydroxycortexone, 17,21-Dihydroxy-4-pregnene-3,20-dione, 17,21-Dihydroxyprogesterone, 4-Pregnene-17α,21-diol-3,20-dione, Cortexolone, Reichstein’s Compound S

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O4
CAS Number:
Molecular Weight:
346.46
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Reichstein′s substance S, ≥98%

Assay

(Infrared Spectrum: Conforms to Structure)
≥98%

form

powder

mp

208 °C (dec.) (lit.)

solubility

ethanol: 50 mg/mL, clear, colorless to faint yellow or tan

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

C[C@]12CCC(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(O)C(=O)CO

InChI

1S/C21H30O4/c1-19-8-5-14(23)11-13(19)3-4-15-16(19)6-9-20(2)17(15)7-10-21(20,25)18(24)12-22/h11,15-17,22,25H,3-10,12H2,1-2H3/t15-,16+,17+,19+,20+,21+/m1/s1

InChI key

WHBHBVVOGNECLV-OBQKJFGGSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In vitro effects of different steroid hormones on superoxide anion production of human neutrophil granulocytes
Gabor Bekesi
Steroids (2000)
E O Yablokov et al.
The Journal of steroid biochemistry and molecular biology, 208, 105793-105793 (2020-12-04)
Steroidogenesis is strictly regulated at multiple levels, as produced steroid hormones are crucial to maintain physiological functions. Cytochrome P450 enzymes are key players in adrenal steroid hormone biosynthesis and function within short redox-chains in mitochondria and endoplasmic reticulum. However, mechanisms
Steven M Weldon et al.
The Journal of pharmacology and experimental therapeutics, 359(1), 142-150 (2016-08-03)
The mineralocorticoid aldosterone is an important regulator of blood pressure, volume, and electrolyte balance. However, excess aldosterone can be deleterious as a driver of vascular remodeling and tissue fibrosis associated with cardiometabolic diseases. Aldosterone synthase (AS) inhibitors (ASI) attenuate the
Masaki Ihara et al.
Analytical chemistry, 81(20), 8298-8304 (2009-09-26)
A noncompetitive immunoassay has the potential for improved sensitivity and working range compared with corresponding competitive assays. However, monovalent antigens with less than 1000 in molecular weight are not susceptible to sandwich assays due to their small size. As a
Rosario Pivonello et al.
The lancet. Diabetes & endocrinology, 8(9), 748-761 (2020-07-31)
Cushing's disease is a rare endocrine disorder characterised by cortisol overproduction with severe complications. Therapies for cortisol reduction are often necessary. Here we report the outcomes from the pivotal phase III study of osilodrostat (a potent oral inhibitor of cytochrome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service