Skip to Content
Merck
All Photos(1)

Key Documents

A5670

Sigma-Aldrich

Anti-Pig IgG (whole molecule)−Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Pig Antibodies, Anti-Pig Antibodies - Anti-Pig IgG (whole molecule)-Peroxidase antibody produced in rabbit, Anti-Porcine IgG (whole molecule)−Peroxidase antibody produced in rabbit, Anti-Swine IgG (whole molecule)−Peroxidase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

pig

technique(s)

direct ELISA: 1:40,000
dot blot: 1:150,000 (chemiluminescent)

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin is a glycoprotein, with two heavy chain and two light chain connected by disulfide bond. IgG is a major class of immunoglobulin. It has five subclasses of IgG- IgG1, IgG3, IgG2a and IgG2b. Affinity isolated antibody is obtained from rabbit anti-pig IgG antiserum by immunospecific purification which removes essentially all rabbit serum proteins, including immunoglobulins, which do not specifically bind to pig IgG. Anti-Pig IgG is then conjugated to peroxidase by protein cross linking with 0.2% glutaraldehyde.

Immunogen

purified pig IgG

Application

Anti-Pig IgG (whole molecule)?Peroxidase antibody produced in rabbit has been used in enzyme-linked immunosorbent assay (ELISA) and immunoblotting.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. It mainly helps in immune defense. Pig IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-pig IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Rabbit anti-pig IgG (whole molecule)-peroxidase antibody binds to pig IgG.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT.

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Livestock susceptibility to infection with Middle East respiratory syndrome coronavirus
Vergara AJ, et al.
Emerging Infectious Diseases, 23(2), 232-232 (2017)
Guy Caljon et al.
PLoS neglected tropical diseases, 8(5), e2911-e2911 (2014-05-24)
Tsetse flies are obligate blood-feeding insects that transmit African trypanosomes responsible for human sleeping sickness and nagana in livestock. The tsetse salivary proteome contains a highly immunogenic family of the endonuclease-like Tsal proteins. In this study, a recombinant version of
Analisis del inmunoblot usando antigeno de cisticercos de Taenia crassiceps en el diagnostico de la cisticercosis
de Arruda PPS, et al.
Biological Chemistry, 56(1-2), 36-42 (2001)
The effect of Trypanosoma evansi infection on pig performance and vaccination against classical swine fever
Holland WG, et al.
Veterinary Parasitology, 111(2-3), 115-123 (2003)
Kristien Van Reeth et al.
Nature communications, 14(1), 7745-7745 (2023-11-27)
Vaccines that protect against any H1N1 influenza A virus strain would be advantageous for use in pigs and humans. Here, we try to induce a pan-H1N1 antibody response in pigs by sequential vaccination with antigenically divergent H1N1 strains. Adjuvanted whole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service