Skip to Content
Merck
All Photos(1)

Key Documents

P8139

Sigma-Aldrich

Phorbol 12-myristate 13-acetate

≥99% (TLC), film or powder, PKC activator

Synonym(s):

12-O-Tetradecanoylphorbol 13-acetate, 4β,9α,12β,13α,20-Pentahydroxytiglia-1,6-dien-3-one 12-tetradecanoate 13-acetate, PMA, TPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H56O8
CAS Number:
Molecular Weight:
616.83
Beilstein:
2407201
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Product Name

Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder

Quality Level

Assay

≥99% (TLC)

form

film or powder

solubility

DMSO: soluble
ethanol: soluble (Dissolve in water-miscible organic solvent prior to preparing aqueous solutions.)

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC(=O)O[C@@H]1[C@@H](C)[C@@]2(O)C(C=C(CO)C[C@@]3(O)C2C=C(C)C3=O)C4C(C)(C)[C@]14OC(C)=O

InChI

1S/C36H56O8/c1-7-8-9-10-11-12-13-14-15-16-17-18-29(39)43-32-24(3)35(42)27(30-33(5,6)36(30,32)44-25(4)38)20-26(22-37)21-34(41)28(35)19-23(2)31(34)40/h19-20,24,27-28,30,32,37,41-42H,7-18,21-22H2,1-6H3/t24-,27+,28-,30-,32-,34-,35-,36-/m1/s1

InChI key

PHEDXBVPIONUQT-RGYGYFBISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dissolve in water-miscible organic solvent prior to preparing aqueous solutions. DMSO solutions can be stored dark at -20 °C for at least six months.
Phorbol 12-myristate 13-acetate (PMA) exhibits high inflammatory properties. It stimulates protein kinase C (PKC) activity. PMA induces the release of proteases, cytokines, and NADPH oxidases, leading to tissue damage or histamine production, and promoting vascular remodeling, redness, heat, and tumor promotion. Additionally, it enhances reactive oxygen species (ROS) production and decreases enzymes involved in ROS detoxification. PMA also inhibits the basal level of superoxide dismutase (SOD), catalase, and glutathione peroxidase.
Phorbol 12-myristate 13-acetate is a phorbol ester. It works as a tumor promoter and is involved in gene transcription, cell growth, differentiation, programmed cell death, immune pathway and receptor desensitization via protein kinase C (PKC) signaling pathways. Phorbol 12-myristate 13-acetate is also a model agent to study growth factors-, hormones-, and cytokines-mediated regulation of cellular growth and differentiation.

Application

Phorbol 12-myristate 13-acetate has been used:
  • to induce differentiation of THP-1monocytes to macrophage.
  • to stimulate the proliferation of peripheral blood lymphocytes.
  • to induce respiratory bursts and enhance the generation of ultra-weak photon emission (UPE) from the neutrophil-like cells derived from HL-60 cells.
  • in conjugation with Ionomycin calcium salt to stimulate T-cell activation
  • for the stimulation of various cells including spleen cells, quiescent lymphocytes, and fetal thymocytes.
  • for the activation of PKC (protein kinase C) in cells.

Features and Benefits

This compound is featured on the PKC page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Caution

Photosensitive

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of protein kinase C results in decreased expression of bovine leukemia virus.
Jensen WA et al.
Journal of Virology, 66, 4427-4427 (1992)
Phorbol 12-Myristate 13-Acetate Induced Toxicity Study and the Role of Tangeretin in Abrogating HIF-1 alpha-NF-kappa B Crosstalk In Vitro and In Vivo
Chang SN, et al.
International Journal of Molecular Sciences (2020)
Cytokine requirements for the growth and development of mouse NK cells in vitro.
Toomey JA et al.
Journal of Leukocyte Biology, 74, 233-233 (2003)
Optical spectral analysis of ultra-weak photon emission from tissue culture and yeast cells
Nerudova M, et al.
Journal of Separation Science, 9450, 162-168 (2015)
Liwang Liu et al.
Journal of biophotonics, 12(8), e201900045-e201900045 (2019-05-31)
Advances in microscopy with new visualization possibilities often bring dramatic progress to our understanding of the intriguing cellular machinery. Picosecond optoacoustic micro-spectroscopy is an optical technique based on ultrafast pump-probe generation and detection of hypersound on time durations of picoseconds

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service