Skip to Content
Merck
All Photos(1)

Key Documents

P5472

Sigma-Aldrich

Precast Agarose Gels

8-well (portrait orientation)

Synonym(s):

Precast agarose gels for high resolution nucleic acids separation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NB.22

grade

for molecular biology

form

gel form

contains

0.5 μg/mL ethidium bromide

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Precast agarose gels are designed for high resolution separation of nucleic acids, producing sharp DNA bands with low background fluorescence. Precast gels are individually foil sealed in plastic trays and are supplied in a box of twenty.

Gel size: 6.0 × 9.5 cm, 5.5 mm thick
Cassette size: 6.8 × 10.2 cm container tray (fits common horizontal electrophoresis units)
Well orientation: 8-well, portrait style

Application

Precast agarose gels are designed for high resolution separation of nucleic acids, producing sharp DNA and RNA bands with low background fluorescence. Precast gels are individually foil sealed in plastic trays that help eliminate contamination from equipment and handling. Gels can be electrophoresed right in the plastic tray.

TBE - agarose gels are available in two common concentrations (1% and 4%) for DNA electrophoresis. Ethidium bromide (0.5 μg/ml) is included in the TBE-agarose gels for easy visualization. TBE-agarose gels are available in 8, 20 and 24 well formats. The separation range for the 1.0% gel is 500-1000 bp and 8-1000 bp for the 4% gel.

MOPS - agarose gels (1.25%) are available without denaturants (e.g. formaldehyde or glyoxal) for the analysis of total RNA, in vitro RNA transcripts, and Northern blotting. RNA electrophoresis usually does not interfere with staining. If significant secondary structure is suspected, the RNA should be denatured by a method listed in the product insert. The separation range for the 1.25% MOPS gel is 0.25-10 kb.
Suitable for separation of DNA fragments ranging from 500 - 1000 base pairs.

Packaging

Package of 20 individually foil-wrapped gels.

Components

Precast gels contain 1.0% agarose in 1X TBE buffer with 0.5 ug/ml ethidium bromide.

Other Notes

DNA agarose gels contain 1× TBE buffer (0.089 M Tris-borate, pH 8.3, 2 mM EDTA) and 0.5 mg/ml ethidium bromide.
RNA agarose gels contain 1× MOPS (20 mM MOPS, 5 mM sodium acetate, 1 mM EDTA and 1 mM EGTA, pH 7.0)

Quality

No detected DNase or RNase activity

Physical form

Gel size: 6.0 × 9.5 cm, 5.5 mm thick
Cassette size: 6.8 × 10.2 cm container tray; fits common horizontal electrophoresis units
Sample format: 8-well (portrait), 20-well (landscape), 24-well (2 × 12 well portrait)

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ian Cheong et al.
Science (New York, N.Y.), 314(5803), 1308-1311 (2006-11-25)
Clostridium novyi-NT is an anaerobic bacterium that can infect hypoxic regions within experimental tumors. Because C. novyi-NT lyses red blood cells, we hypothesized that its membrane-disrupting properties could be exploited to enhance the release of liposome-encapsulated drugs within tumors. Here
R Kaminski et al.
Gene therapy, 23(8-9), 690-695 (2016-05-20)
A CRISPR/Cas9 gene editing strategy has been remarkable in excising segments of integrated HIV-1 DNA sequences from the genome of latently infected human cell lines and by introducing InDel mutations, suppressing HIV-1 replication in patient-derived CD4+ T-cells, ex vivo. Here

Articles

Colony PCR reagents and our colony PCR protocol using REDExtract-N-Amp™ PCR ReadyMix™ and JumpStart™ REDTaq® PCR ReadyMix™ reagents.

Protocols

These gels are suitable for separating nucleic acids, giving sharp DNA bands and low background fluorescence.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service