Skip to Content
Merck
All Photos(1)

Key Documents

M5755

Sigma-Aldrich

MOPS-EDTA-Sodium Acetate (MESA)

powder for RNA electrophoresis buffers

Synonym(s):

MESA Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25

grade

for molecular biology

product line

BioReagent

form

powder

technique(s)

electrophoresis: suitable

pH

6.8-7.2(reconstituted per directions)

solubility

H2O: soluble, faintly yellow

foreign activity

DNAses, RNases, NICKases, endonucleases, and exonucleases, none detected

storage temp.

room temp

General description

MOPS – EDTA – Sodium Acetate (MESA) is the most commonly used running buffer for RNA electrophoresis prior to Northern blotting procedures. Applied voltage of 5 V/cm is recommended for maximum resolution.

Application

Suitable for use when making formaldehyde-agarose gels and associated running buffer for RNA electrophoresis.

Components

1x MESA buffer contains:
  • 40 mM MOPS
  • 10 mM sodium acetate
  • 1 mM EDTA (pH 8.3)

Reconstitution

Dissolve entire contents of bottle in a final volume of 1L molecular biology grade water. This produces a 10x MESA stock solution that can be further diluted as needed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sambrook, J., et al.
Molecular Cloning: A Laboratory Manual, 7-7 (1989)
Molecular techniques for detection of gene expression in neuroimmunology.
Cowan, E.P., and Dhib-Jalbut, S.S.
Methods in Neuroscience, 24, 41-41 (1995)
Kathryn M Jones et al.
Journal of bacteriology, 184(9), 2491-2499 (2002-04-12)
Two operons have been cloned from Anabaena sp. strain PCC 7120 DNA, each of which encodes the three core subunits of distinct mitochondrial-type cytochrome c oxidases. The two operons are only 72 to 85% similar to one another at the
Xueyu Xiang et al.
Developmental biology, 275(2), 343-355 (2004-10-27)
Recently, we cloned and sequenced the cDNA of allurin, a sperm chemoattractant isolated from the jelly of Xenopus laevis eggs [Proc. Natl. Acad. Sci. U.S.A. 78 (2001) 11205]. In this report, we demonstrate that allurin mRNA is expressed almost exclusively
Sheetal Sharma et al.
Cell reports, 34(1), 108565-108565 (2021-01-07)
The MRE11-RAD50-NBS1 (MRN) complex supports the synthesis of damage-induced long non-coding RNA (dilncRNA) by RNA polymerase II (RNAPII) from DNA double-strand breaks (DSBs) by an unknown mechanism. Here, we show that recombinant human MRN and native RNAPII are sufficient to

Protocols

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

TAE and TBE are both used as running buffers for nucleic acid electrophoresis but have some important differences. Review our recipes and video to give your application the best chance of success.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service