Skip to Content
Merck
All Photos(5)

Key Documents

H7033

Sigma-Aldrich

PerfectHyb Plus Hybridization Buffer

for Northern and Southern blotting, solution

Synonym(s):

hyb buffer, hyb solution, hybridization solution, northern hybridization buffer, southern hybridization buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.25
Pricing and availability is not currently available.

General description

Sigma′s PerfectHyb Plus is a ready-to-use hybridization buffer for Northern and Southern blot protocols. Use of PerfectHyb Plus can decrease hybridization time to as little as two hours.

Application

Suitable for use in hybridization buffers used in Northern blot, Southern blot, colony/plaque blots, and dot blots.

Features and Benefits

  • Fast and easy-to-use
  • Up to 20x more sensitive than conventional buffers
  • Doesn′t require blocking steps
  • Compatible with neutral and positively-charged membranes
  • Compatible with chemiluminescent, colorimetric , and DIG detection assays
  • Compatible with radioactive and non-radioactive probes
  • Compatible with single or double-stranded DNA, RNA, and oligonucleotide probes

Other Notes

A small amount of precipitate that has formed in the solution can be dissolved at 30-35°C with occasional gentle inversion.

Legal Information

PerfectHyb is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alavala Matta Reddy et al.
BMC genomics, 10, 65-65 (2009-02-07)
MicroRNAs (miRNAs) are small ~22-nt regulatory RNAs that can silence target genes, by blocking their protein production or degrading the mRNAs. Pig is an important animal in the agriculture industry because of its utility in the meat production. Besides, pig
Yinping Jiao et al.
BMC plant biology, 11, 141-141 (2011-10-22)
miRNAs are known to play important regulatory roles throughout plant development. Until recently, nearly all the miRNAs in maize were identified by comparative analysis to miRNAs sequences of other plant species, such as rice and Arabidopsis. To find new miRNA
Edyta Koscianska et al.
TheScientificWorldJournal, 11, 102-117 (2011-01-11)
This protocol describes how to perform northern blot analyses to detect microRNAs and their precursors with single-nucleotide resolution, which is crucial for analyzing individual length variants and for evaluating relative quantities of unique microRNAs in cells. Northern blot analysis consists
Angel Emilio Martínez de Alba et al.
Nucleic acids research, 39(21), 9339-9344 (2011-08-05)
In plants, most microRNAs (miRNAs) and several endogenous small interfering RNAs (siRNAs) bind to ARGONAUTE1 (AGO1) to regulate the expression of endogenous genes through post-transcriptional gene silencing (PTGS). AGO1 also participates in a siRNA-mediated PTGS defense response that thwarts exogenous
Shinpei Kawaoka et al.
RNA (New York, N.Y.), 15(7), 1258-1264 (2009-05-23)
Genetic studies and large-scale sequencing experiments have revealed that the PIWI subfamily proteins and PIWI-interacting RNAs (piRNAs) play an important role in germ line development and transposon control. Biochemical studies in vitro have greatly contributed to the understanding of small

Protocols

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service